Learn More
Creative Biomart Interferon Tau (IFN-Tau)
Supplier:  Creative Biomart THP0270
Encompass_Preferred
IFN-Tau Ovine Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 172 amino acids and having a molecular mass of 19914.7 Dalton. The IFN-Tau is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4321
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.