Learn More
BINDER™ Interior Lighting
SDP
Catalog No. 502875704 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 130, KB 260, KB 470
KB 1600
KB 65
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 1600
KB PRO 260/470
KB PRO 720, KB PRO 1060
KBF 130, KBF 260, KBF 470
KBF 1600
KBF 720, KBF 1060
KBF PRO 1600
KBF PRO 720, KBF PRO 1060
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5704 KB 65
50-287-5676 KB 130, KB 260, KB 470
50-287-5765 KB PRO 130/260/470
50-287-5614 KBF 130, KBF 260, KBF 470
50-287-5635 KB PRO 260/470
50-287-5660 KB 720, KB 1060
50-287-5766 KB PRO 720, KB PRO 1060
50-287-5767 KBF 720, KBF 1060
50-287-5720 KBF PRO 720, KBF PRO 1060
50-287-5700 KB PRO 1600
50-287-5689 KBF 1600
50-287-5594 KBF PRO 1600
50-287-5655 KB 1600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 50-287-5704 Supplier BINDER™ Supplier No. 80122555
Only null left
Add to Cart
Add to Cart
Interior lighting with climate-resistant LEDs.
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Specifications
For Use With (Equipment) KB 65
Material Glass
Type Lighting