Learn More
STEMCELL Technologies Mouse Recombinant IL-13, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78030
Encompass_Preferred
Interleukin 13 (IL-13) is a type I cytokine, and signals through type I cytokine receptors to activate JAK/STAT and IRS-1/IRS-2 pathways. IL-13 is produced by T cells and innate lymphoid cells (Pulendran & Artis) and it inhibits production of pro-inflammatory cytokines, prostaglandins, and reactive oxygen and nitrogen species by monocytes and macrophages (Hershey). Unlike human B cells, IL-13 has no effect on mouse B cell development and function. IL-13 promotes eosinophil survival, activation, and recruitment, and also activates mast cells (Hershey). IL-13 regulates gastrointestinal parasite expulsion, airway hyperresponsiveness, allergic inflammation, tissue eosinophilia, goblet cell hyperplasia, intracellular parasitism, tissue remodeling, tumor cell growth, and fibrosis. IL-13 induces hypercontractility of smooth muscles by directly acting on the muscle cells as well as the enteric nerves (Wynn).
Catalog No. 50-226-9604
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.