Learn More
Interlocked™ Vial with Fused Glass Insert, Sold by MilliporeSigma™ Supelco™
Catalog No. 111010566 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Screw Thread Vial
Snap Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
11-101-0566 Screw Thread Vial
11-101-0599 Snap Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-0566 Supplier MilliporeSigma Supelco Supplier No. 29109U
Add to Cart
Add to Cart
A 300 muL flange top glass insert permanently fused into 12 x 32 mm glass vials.
  • The new Interlock Vial/Insert limited volume vial is a 300 muL flange top glass insert permanently fused into 12 x 32 mm glass vials.
  • This product provides the user with the convenience and performance of a one piece microvial at a fraction of the cost.
  • The flange top centers the inserts into the vial.
  • Glass to glass fusion eliminates concerns about contaminants.
  • Vials are made of borosilicate, Type 1 glass with a choice of clear or amber glass.
TRUSTED_SUSTAINABILITY
Specifications
Material Clear Glass
Volume (Metric) 300 L
Quantity 100/Pk.
Product Type Screw Thread Vial
Interlocked is a trademark of J.G. Finneran Associates, Inc.