Learn More
Argos Technologies™ Interlocking Glove Box Storage Rack
Catalog No. 03391566 Shop All Oakton Products
Change view
Click to view available options
Single
Catalog No.
03-391-566 Single
1 options
This item is not returnable. View return policy
Catalog No. 03-391-566 Supplier Argos Technologies™ Supplier No. 0439699
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Epoxy-coated steel glove rack provides complete laboratory flexibility and is compatible with most major brands of disposable glove boxes.
  • Epoxy-coated steel glove rack provides complete laboratory flexibility
  • Coated wire stopper prevents glove boxes from falling out
  • Easily slides together to accommodate additional racks
  • Racks can be mounted horizontally or vertically
  • Mounting hardware included
Specifications
Product Type Glove Box Holder
Color Blue
Dimensions (L x W x H) 10.25 x 7 x 4 in.
Holds Disposable Glove Boxes
Material Epoxy-coated Steel
Mounting Keyhole Slots
No. of Compartments 1
Includes Mounting hardware