Learn More
Carson™ Intermediate 100 to 1000x Biological Compound Microscope with Mechanical Stage
SDPSCI_ED
Catalog No. S81733 Shop All Carson Optical Products
Change view
Click to view available options
Head Style:
Monocular
Catalog No. Head Style
S81733 Monocular
1 options
Catalog No. S81733 Supplier Carson™ Supplier No. MS100
Add to Cart
Add to Cart
Intermediate table-top microscope for high school/college students, hobbyists or technicians, metal body/monocular head at 45°
  • Monocular head rotates 360° for easy sharing of specimen view
  • Coarse and fine focus knobs to help user to achieve a sharp focus
  • LED illumination provides bright white colored light and intensity control to set optimal lighting conditions
  • Backed by Carson™'s one year limited warranty
  • Includes:
    • Flter holder with included blue color filter
    • AC power adaptor for use internationally without a voltage transformer
  • Specifications
    Eyepiece WF 10X
    Head Style Monocular
    Lighting Type LED Upper and Lower
    Magnification Power 100X to 1000X
    Model MS-100
    Product Type Double layer, Condenser: Abbe 1.25 NA with iris diaphragm filter
    For Use With (Application) Student microscope
    Includes AC power adapter, Filter holder with included blue color filter
    Material Aluminum