Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Intermediate Density Lipoproteins Native Protein
SDP

Catalog No. N19953910U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N19953910U 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N19953910U Supplier Novus Biologicals™ Supplier No. NBP1995391000UG

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page

Native Intermediate Density Lipoproteins

The Intermediate Density Lipoproteins Native Protein is derived from Human Plasma. The Intermediate Density Lipoproteins Native Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Gene ID (Entrez) 4035
Molecular Weight (g/mol) 5002.5 kDa
Quantity 1000 μg
Source Human Plasma
Storage Requirements Store at 4°C.
Regulatory Status RUO
Specific Reactivity Intermediate Density Lipoproteins
Conjugate Unconjugated
Species Human
Buffer 150 mM NaCl, 0.01% EDTA, pH 7.4
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.