Learn More
United Scientific™ Intermediate Spectrometer
SDPSCI_ED
Catalog No. S139468 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Intermediate Spectrometer
Catalog No. Type
S139468 Intermediate Spectrometer
1 options
This item is not returnable. View return policy
Catalog No. S139468 Supplier United Scientific™ Supplier No. SPECT02
Add to Cart
Add to Cart
This item is not returnable. View return policy
Spectrometer that is easy to use and easy to set up.
This instrument is easy to set up, easy to use, and is designed to make precise measurements.
  • Features high quality 22 mm optics, a built-in illuminated auto-collimator for fast set up, a 17.8 cm diameter divided circle with clear graduations, a double-ended vernier with illuminated magnifier reading to 1 ft. resolution, and a 7 cm diameter prism table which rotates 360° and adjusts 2 cm in height to accommodate various accessories.
  • The spectrometer is supplied with diffraction grating and grating holder, prism, illuminated magnifier, auto-collimation mirror, power supply (for 110 VAC), and an instruction manual.
  • Additional Spectrometer Prisms and Diffraction Gratings are available separately.
  • Shipping weight: 29 lb.
  • Dimensions: 17 x 17 x 12 in.
Specifications
Type Intermediate Spectrometer
Prop 65 Warning: Cancer and reproductive harm