Learn More
United Scientific™ Intermediate Spectrometer with 30 mm Optics
SDPSCI_ED
Catalog No. S139471 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Intermediate Spectrometer
Catalog No. Type
S139471 Intermediate Spectrometer
1 options
This item is not returnable. View return policy
Catalog No. S139471 Supplier United Scientific™ Supplier No. SPECT03
Add to Cart
Add to Cart
This item is not returnable. View return policy
Spectrometer that provides brighter line images and makes precise measurements easier.
  • This instrument is an enhanced version of our popular Intermediate Spectrometer (SPECT02.)
  • Its larger 30 mm diameter optics give brighter line images and make precise measurements easier.
  • The optical focusing system has also been refined and the instrument retains the same premium features as the earlier version, including the built-in illuminated autocollimator for fast set up, 17.8 cm diameter divided circle, double-ended vernier reading to 1 ft. resolution, and 7 cm diameter rotatable and height-adjusting prism table to accommodate various accessories.
  • The spectrometer is supplied with a 300 lines/mm diffraction grating and grating holder, high quality prism, illuminated magnifier, auto-collimation mirror with a power supply for 110 VAC, and an instruction manual.
  • Additional Spectrometer Prisms and Diffraction Gratings are available separately.
Specifications
Type Intermediate Spectrometer
Prop 65 Warning: Cancer and reproductive harm