Learn More
CPC Scientific H-Pro-His-Ala-Gln-Leu-Leu-Arg-Val-Gly-Cys-Val-Leu-Gly-Thr-Cys-Gln-Val-Gln-Asn-Leu-Ser-His-Arg-Leu-Trp-Gln-Leu-Val-Arg-Pro-Ser-Gly-Arg-Arg-Asp-Ser-Ala-Pro-Val-Asp-Pro-Ser-Ser-Pro-His-Ser-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HITM002A
Encompass
Intermedins are members of the calcitonin/CGRP family. In vitro studies have shown that the synthetic 47 amino acid intermedin peptides act through the calcitonin receptor-like receptor/receptor activity-modifying protein (CRLR/RAMP) complexes by increasing intracellular cAMP levels. Unlike calcitonin gene-related peptide (CGRP) and adrenomedullin (ADM) intermedin exhibited no preference for one of the three RAMPs when co-expressed with CRLR. In normal and spontaneously hypertensive rats it has been demonstrated that intermedin treatment results in blood pressure reduction which can be blocked by the CGRP receptor antagonist CGRP (8-37). Apart from the hypotensive action, synthetic intermedin has also been shown to inhibit gastric emptying and food intake in mice. Therefore, intermedin represents an additional regulator of gastrointestinal and cardiovascular functions and might be involved in other bioactivities mediated by the CRLR/RAMP receptor complexes. ONE-LETTER SEQUENCE: PHAQL
Catalog No. 50-210-9529
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.