Learn More
BIOHIPPO Internal Ctrl Lentivirus LV
Supplier: BIOHIPPO BHV194000427
Internal Ctrl Lentivirus is a transcription factor reporter lentiviral vector designed to monitor the Estrogen receptor (ER) signaling pathway signaling pathway in living cells It delivers a Renilla Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The lentiviral vector pseudotyped with VSV-G enables efficient gene delivery and stable genomic integration Typical applications include pathway activity monitoring assay development high content screening and mechanism of action studies It is suitable for use with Human/mouse cells This product is intended for research use only and is not for diagnostic or therapeutic procedures