Learn More
Eisco™ Internal Gear and Pinion Drive Mechanical Demonstration Model
SCI_ED
Catalog No. S27667 Shop All Eisco Products
Change view
Click to view available options
Internal Gear and Pinion Drive Mechanical Demonstration Model
Catalog No.
S27667 Internal Gear and Pinion Drive Mechanical Demonstration Model
1 options
Catalog No. S27667 Supplier Eisco™ Supplier No. PH0366TM13
Add to Cart
Add to Cart
The Internal Gear and Pinion Drive is an interactive visual demonstration of a compact and efficient gear mechansism design affording high torque and gear reduction capabilities.
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how an internal Gear and Pinion drive works.
  • Two dials to show rotation.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Product Type Internal Gear and Pinion Drive Mechanical Demonstration Model