Learn More
International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Boot Covers
Shop All International Enviroguard Systems ProductsDescription
These microporous boot covers are clean processed to offer the perfect balance of safety and comfort. The fabric keeps dust and liquids out while allowing air to flow freely, so your feet stay cool throughout a hard day's work. These disposable boot covers are ideal for non-hazardous work environments, such as controlled environments or cleanrooms, where you need reliable liquid and particulate protection. Boot covers are designed with tunnelized elastic to contain particle shed. Non-skid soles provide added traction to help prevent accidental slips and trips. Each boot cover is individually packaged to help prevent cross-contamination.
- Non-skid soles help improve traction to prevent accidental slips
- Elastic closures for snug, secure fit that keeps boot covers in place
- Available in Bulk or Individually packaged to prevent cross-contamination
Specifications
Specifications
| Size | X-Large |
| Material | Microporous Fabric |
| Color | White |
| Closure Type | Elastic Top and Ankles |
| Sole Type | Non-skid |
| Seam Type | Serged |
| Thickness | 29 mil |
| Certifications/Compliance | ANSI ISEA 101-1996 R2020, ASTM T903, ASTM F1670-07 |
| Packaging Quantity | 200/Cs. |
| Packaging Type | Bulk Packaged |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.