Learn More
International Enviroguard™ MicroGuard CE™ Cleanroom Frocks
Shop All International Enviroguard Systems ProductsDescription
- Microporous, clean processed frocks available in individual or bulk packaging
- Fabric keeps dust and liquids out while allowing air to flow freely
- Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
- Designed with tunnelized elastic to contain particle shed
- Bound mandarin collar for enhanced protection
- Front snap closures are easy to secure
- Constructed with bound seams to eliminate raw edges and strengthen the seam
- No pockets
Specifications
Specifications
| Size | Large |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Collar Type | Bound Mandarin |
| Size (English) Chest | 40 in. |
| Size (Metric) Chest | 101.6 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.