Learn More
International Enviroguard™ MicroGuard CE™ Cleanroom Sleeves
Shop All International Enviroguard Systems ProductsDescription
- Microporous, clean processed sleeves
- Fabric keeps dust and liquids out while allowing air to flow freely
- Ideal for nonhazardous work environments to protect against liquid and particulates
- Designed with tunnelized elastic to offer a more secure closure, stronger seams, and the elimination of raw edges that may generate unwanted particulates
- Each sleeve is individually packaged to help prevent cross-contamination
- Elastic closures on both ends of sleeves for secure fit
- Environmentally friendly - can be recycled after use
- 18 in. length provides extended arm coverage
Order Info
Available in bulk packages or individually wrapped clean process packages
Specifications
Specifications
| Product Type | Controlled Environment Sleeves |
| Size | Standard |
| Material | Microporous Fabric |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Cuff Style | Elastic |
| Model | CE8065 |
| Length (English) | 18 in. |
| Length (Metric) | 45.7 cm |
| Packaging | Bulk Packaged |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.