Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Coveralls

Catalog No. 19472511 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging:
Bulk
Individually Packaged
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging
19-472-511 3X-Large Individually Packaged
19-472-507 Medium Individually Packaged
19-472-512 4X-Large Individually Packaged
19-472-829 X-Large Individually Packaged
19-472-502 Large Bulk
19-472-501 Medium Bulk
19-472-503 X-Large Bulk
19-472-504 2X-Large Bulk
19-472-505 3X-Large Bulk
19-472-506 4X-Large Bulk
19-472-509 X-Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-472-511 Supplier International Enviroguard™ Supplier No. CE8013CI3XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Clean processed microporous coveralls offer the perfect balance of safety and comfort. Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed.

These clean processed coveralls offer the perfect balance of safety and comfort. Microporous fabric keeps dust and liquids out while allowing air to flow freely, so you stay cool as you put in a hard day's work. These coveralls are ideal for non-hazardous work environments where you need reliable liquid and particulate protection. Coveralls are designed with tunnelized elastic to offer a more secure closure, stronger seams, and the elimination of raw edges that may generate unwanted particulates. Each coverall is individually packaged to help prevent cross-contamination.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area.
  • Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Secure elastic wrists and ankles help keep contaminants out.
  • Environmentally friendly - can be recycled after use.

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Model MicroGuard CE™
No. of Pockets None
Size (English) Chest 45 in.
Size (English) Inseam 32 in.
Size (English) Waist 42 in.
Size (Metric) Chest 114.3 cm
Size (Metric) Inseam 81.28 cm
Size (Metric) Waist 106.6 cm
For Use With (Application) Liquid barrier and particulate barrier in controlled environments
Packaging Individually Packaged
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.