Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Coveralls with Attached Hood and Boots

Catalog No. 19472515 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging:
Bulk
Individually Packaged
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging
19-472-515 X-Large Bulk
19-472-519 Medium Individually Packaged
19-472-520 Large Individually Packaged
19-472-521 X-Large Individually Packaged
19-472-523 3X-Large Individually Packaged
19-472-522 2X-Large Individually Packaged
19-472-524 4X-Large Individually Packaged
19-472-513 Medium Bulk
19-472-514 Large Bulk
19-472-518 4X-Large Bulk
19-472-516 2X-Large Bulk
19-472-517 3X-Large Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-472-515 Supplier International Enviroguard™ Supplier No. CE8019BPXL
This item is not returnable. View return policy

Clean processed coveralls are made from microporous fabric to protect against dust and liquids while allowing air to flow freely.

  • Clean processed coveralls offer the balance of safety and comfort. Microporous fabric keeps dust and liquids out while allowing air to flow freely, so you stay cool as you put in a hard day's work. Ideal for non-hazardous work environments where you need reliable liquid and particulate protection. Coveralls are designed with tunnelized elastic to contain particle shed and eliminate raw edges that may generate unwanted particulates. With an attached hood and boots, you have full-body protection from head-to-toe.
  • Packaging
    • Bulk
    • Individually Packaged: Each coverall is individually packaged to help prevent cross-contamination
  • Attached hood and boots provide more complete body protection
  • Roomy, seamless shoulder area resists tears in high-movement areas
  • Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric
  • Tunnelized elastic back stretches with the user for more comfort and higher tear resistance
  • Secure elastic wrists and ankles help keep sleeves and pant legs in place to prevent skin exposure
  • Environmentally friendly - can be recycled after use

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Model MicroGuard CE™
Includes Attached Hood and Boots
Size (English) Chest 42 in.
Size (English) Inseam 30 in.
Size (English) Waist 39 in.
Size (Metric) Chest 106.6 cm
Size (Metric) Inseam 76.2 cm
Size (Metric) Waist 99.06 cm
For Use With (Application) Liquid barrier and particulate barrier in controlled environments
Packaging Bulk
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.