Learn More
International Enviroguard™ MicroGuard CE™ Coveralls with Attached Hood and Boots
Shop All International Enviroguard Systems ProductsDescription
- Clean processed coveralls offer the balance of safety and comfort. Microporous fabric keeps dust and liquids out while allowing air to flow freely, so you stay cool as you put in a hard day's work. Ideal for non-hazardous work environments where you need reliable liquid and particulate protection. Coveralls are designed with tunnelized elastic to contain particle shed and eliminate raw edges that may generate unwanted particulates. With an attached hood and boots, you have full-body protection from head-to-toe.
- Packaging
- Bulk
- Individually Packaged: Each coverall is individually packaged to help prevent cross-contamination
- Attached hood and boots provide more complete body protection
- Roomy, seamless shoulder area resists tears in high-movement areas
- Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric
- Tunnelized elastic back stretches with the user for more comfort and higher tear resistance
- Secure elastic wrists and ankles help keep sleeves and pant legs in place to prevent skin exposure
- Environmentally friendly - can be recycled after use
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Large |
| Material | Microporous Composite Fabric |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic, Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Attached |
| Closure Type | Zipper |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.