Learn More
International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Shoe Covers
Shop All International Enviroguard Systems ProductsDescription
- Clean-processed shoe covers provide protection against particulates and non-hazardous liquids
- Breathable fabric allows moisture vapor to escape to help keep feet cooler
- Tunnelized elastic at the top contains particle shed and eliminates raw edges
- Environmentally friendly and can be recycled after use
- Non-skid soles help improve traction to prevent accidental slips
- Elastic closures for snug, secure fit that keeps boot covers in place
- Individually packaged
Specifications
Specifications
| Size | Large |
| Material | Microporous Fabric |
| Color | White |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Non-skid |
| Seam Type | Serged |
| Thickness | 20 mil |
| Certifications/Compliance | ANSI ISEA 101-1996 R2020, ASTM T903, ASTM 1670-07 |
| Packaging Quantity | 200/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.