Learn More
Invitrogen™ eBioscience™ Intracellular Fixation & Permeabilization Buffer (plus Brefeldin A)
Catalog No. 501128915
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-112-8915 1 kit
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-8915 Supplier Invitrogen™ Supplier No. 88882388
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart
Contains Sodium azide and formaldehyde
Includes
  • Brefeldin A solution
  • IC Fixation Buffer
  • Permeabilization Buffer (10X) . Permeabilization Buffer is supplied as a 10X concentrate. 1X Permeabilization Buffer contains 0.1% saponin and 0.09% sodium azide.
Designed for use in preparation of living cells for intracellular staining and flow cytometric analysis. A protein transport inhibitor (Brefeldin A) is included for treatment of cells during culture, to cause accumulation of cytokine protein at the Golgi and enhance resulting staining.
  • Fixation buffer is used to cross-link, stabilize, and fix the cell membrane
  • Permeabilization buffer is used to reversibly permeabilize the cell membrane so the staining antibodies can enter the cell effectively
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Fixation Reagent, Permeabilization Reagent
Product Line eBioscience
Product Type Fixation/Permeabilization Buffer
For Research Use Only. Not for use in diagnostic procedures.