Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ IC Fixation Buffer

Catalog No. 501129058
Encompass_Preferred
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9058 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains formaldehyde
Catalog No. 50-112-9058 Supplier Invitrogen™ Supplier No. 00822249
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Contains formaldehyde

Can be used to fix cells prior to permeabilization with the Permeabilization Buffer (cat. 00-8333) when performing intracellular staining of antigens located in the cytoplasm and secretory pathway, such as cytokines, chemokines and growth factors.

  • Compatible with surface-staining fluorochrome-conjugated antibodies
  • Can be used as a short-term storage buffer for fluorochrome (including tandem dyes)-stained cells for up to 3 days at 2° to 8°C (stored in the dark)
TRUSTED_SUSTAINABILITY

Specifications

Quantity 125 mL
Reagent Type Fixation Reagent
Product Line eBioscience
Product Type Fixation Buffer
Can I use homemade fixation buffers (or other commercially available fixatives) on my cells after staining with eVolve-conjugated antibodies?

No. For optimal results, we recommend that you use the IC Fixation Buffer (Cat. No. 00-8222-49) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54 or 00-5333-57). Impurities in fixation buffers can severely affect fluorochrome brightness.

Will eVolve fluorophores survive fixation?

Absolutely. eVolve fluorophores retain their fluorescent properties after fixation with eBioscience's fixation buffers such as IC Fixation Buffer (Cat. No. 00-8222-49).

Can I use homemade fixation buffers (or other commercially available fixative) on my cells followed by flow cytometric analysis?

No. We do not recommend the use of other fixatives. We validate our studies using eBioscience's IC Fixation Buffer (Cat. Nos. 00-8222-49, 00-8222-52) and 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Other recipes or manufacturer's products may give inferior results.

I can not run my samples for flow cytometric analysis immediately. What do I do to preserve the best intracellular staining?

For optimal results, we recommend that you analyze stained cells immediately.

However, if you do need to store your samples that were stained with antibodies conjugated to organic fluorochromes, we recommend you complete your staining protocol and fix your samples with IC Fixation Buffer (Cat. Nos. 00-8222-49, 00-8222-52) (100 µL sample with 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Cells can be stored in these buffers for up to 3 days in the dark at 4 degrees C.

We have observed minimal impact on brightness or FRET efficiency/compensation when using the IC Fixation (Cat. Nos. 00-8222-49, 00-8222-52) or 1-step Fix/Lyse Solution (Cat. Nos. 00-5333-54, 00-5333-57). Differences in fixation buffer quality can affect fluorochrome brightness or FRET efficiency. Fixation of tandem dyes, such as APC-eFluor 780 and PE-Cy7, does not increase the amount of compensation required from the APC or PE detector, respectively. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

Can samples stained with antibodies conjugates to tandem dyes (i.e., APC-eFluor 780, PE-Cy7, etc.) be fixed and stored?

Yes, samples that are stained with tandem dye conjugated antibodies can be stored in IC Fixation Buffer (Cat. No. 00-8222-49, 00-8222-52) (100 µL sample with 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) for up to 3 days in the dark at 4 degrees C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.