Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Intracellular pH Assay Kit, Cell-based, Fluorescent, 1000 Tests

Catalog No. 502587023
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-7023 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-7023 Supplier Abcam Supplier No. AB2285521000TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative cell-based fluorescent assay kit for measuring intracellular pH in suspension and adherent cells.

  • Assay type: Cell-based
  • Detection method: Fluorescent
  • Results type: Quantitative
  • Sample types: Suspension cells and Adherent cells
  • Recommended for use with Microplate reader
  • Cited in 5 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 1000 Tests
Target Molecule pH
Detection Location In-Cell Detection
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 1000 Tests
For Use With (Application) Intracellular pH assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.