Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Intracellular TE Nano-Glo Substrate/Inhibitor, 10,000 assays

Catalog No. PRN2161 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN2161 10,000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN2161 Supplier Promega Supplier No. N2161
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Intracellular TE Nano-Glo Substrate/Inhibitor is a supplement for NanoBRET™ Target Engagement Assays.

  • Dedicated Reagents for Intracellular NanoBRET™ TE Assays
  • Combination of NanoLuc(R) Substrate and Extracellular NanoLuc(R) Inhibitor
  • Ensures measurements originate from inside intact cells
  • For use with NanoBRET™ Intracellular Target Engagment Tracers

Specifications

Content And Storage -30°C to -10°C
Quantity 10,000 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.