Learn More
BioMed Diagnostics With 10% horse blood, this self-contained system aids in the culture, isolation and identification of fastidious pathogenic bacteria including Taylorella equigenitalis. | Available in: 20 pack
Supplier: BioMed Diagnostics 12133002
With 10% horse blood, this self-contained system aids in the culture, isolation and identification of fastidious pathogenic bacteria including Taylorella equigenitalis. | Product Principle: 1.) Combine collection, culture, result and microscopic observation in one IVD device. 2.) Highly selective prepared media in a device designed for in-vitro specimen transport and incubation. 3.) Rapid presumptive-positive diagnosis without re-plating. Intray can be placed directly on microscope stage. 4.) Fully enclosed cassette with Tyvek air filtration prevents contamination and exposure, post-collection. | Detects: Taylorella equigenitalis | Time to Result: 18-24 hours, incubate at 37C | Available in: 20 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.