Learn More
BioMed Diagnostics Suitable for isolation and easy chromogenic differentiation of L. monocytogenes; provides initial differentiation and presumptive identification. | Available in: 20 pack
Supplier: BioMed Diagnostics 11193002
Suitable for isolation and easy chromogenic differentiation of L. monocytogenes; provides initial differentiation and presumptive identification. COLOREX Listeria and COLOREX ID Listeria, traditionally used in tandem, isolate and chromogenically differentiate colonies confirming the presence of Listeria monocytogenes with prepared plated culture media. When used together, COLOREX Listeria provides initial differentiation and presumptive identification, while COLOREX ID Listeria provides confirmation of suspect L. monocytogenes colonies re-plated from COLOREX Listeria. | Detects: Listeria monocytogenesand L. innocua | Time to Result: 18-24 hours, incubate at 37C. | Available in: 20 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.