Learn More
BioMed Diagnostics Used to aid in the detection of dermatophytes from specimens with mixed microbiota. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, Microsporum gypseum. | Available in: 5 pack
Supplier:  BioMed Diagnostics 11593001
Encompass_Preferred
InTray DM-FungID prepared culture media serves as a microbiology sample collection, transport, and culture device. Its enriched selective medium is formulated to produce distinctive colony growth from clinical specimens with mixed microbiota. Engineered to be selective for positive growth of dermatophytes strains all while inhibiting the growth of both gram+ and gram bacteria, yeast and saprophytic fungi. Selective for Tinea and ringworm commonly used to describe dermatophytes. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: Incubate inoculated trays for up to 14 days at 18-30C, in the dark. Observe the trays daily for color changes through the clear viewing window. | Available in: 5 pack
Catalog No. NC2582846
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.