Learn More
BioMed Diagnostics A nutrient-rich medium for a broad-spectrum of yeasts and molds. | Detects: Yeasts and Molds. | Available in: 5 pack
Supplier:  BioMed Diagnostics 11333001
Encompass_Preferred
InTray Malt Extract allows for simultaneous growth and observation of yeasts and molds. The agar used in this device is recommended for use with clinical specimens, foodstuffs, and cosmetics as well as samples derived from the membrane filtration procedure. | Detects: Yeasts and Molds | Time to Result: Incubate at 25-35C for 18-24 hours or under ambient atmosphere. | Available in: 5 pack
Catalog No. NC2582934
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.