Learn More
BioMed Diagnostics A medium engineered for growth and detection of dermatophyte fungi. | Detects: Trichophyton mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Available in: 20 pack
Supplier:  BioMed Diagnostics 11283002
Encompass_Preferred
Simplify your classical dermatophyte culture and microscopy methods with InTrays FungID device sytem. The prepared plated culture media in InTray FungID SAB w/CC enables simultaneous detection and observation of multiple dermatophyte fungi a specific group of fungi that cause common skin, nail and hair infections in both humans and animals. Dermatophytes are zoonotic, meaning they can be transmitted across species and generally made up of three genera: Microsporum, Epidermophyton and Tyichophyton. | Detects: Trichophyton mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: 1 - 21 days, incubated at 25C for positive dermatophyte identification. | Available in: 20 pack
Catalog No. NC2582965
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.