Learn More
BioMed Diagnostics A self-contained system with an improved formulation including selective antibiotics for isolation and identification of T. equigenitalis. | Available in: 20 pack
Supplier:  BioMed Diagnostics 12143002
Encompass_Preferred
For the isolation of Taylorella equigenitalis. The agar in this device contains an improved formulation with selective antibiotics for the suppression of competing organisms that may be present during sample collection. It is recommended for use in conjunction with a non-selective medium, such as the InTray Chocolate Eugon. | Detects: Trichophyton mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: 18-24 hours, incubate at 37C | Available in: 20 pack
Catalog No. NC2582966
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.