Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Introductory Optical Bench Set
SDPSCI_ED

Catalog No. S43682 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43682 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43682 Supplier United Scientific Supplies Supplier No. OBSET4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An economical, high quality bench set that works well for introductory lens work.

  • This introductory bench set allows the basic properties of lenses to be explored.
  • The optical elements are mounted in robust molded holders that are pushed into sockets in the riders.
  • Each rider can accept up to three elements.
  • The riders slide freely on the aluminum bench until an optical element is pushed in, when the rider locks in place. This eliminates accidental repositioning allowing the entire lightweight setup to be carried around without anything falling off.
  • The LED-illuminated object is self-contained (powered by an AAA battery– no cables) and generates bright, clear images on the opaque screen.
  • Four high quality lenses are included, all with 50 mm diameter.
  • The focal lengths are, +100 mm, +150 mm, +200 mm, (all convex) and– 150 mm (concave).

Specifications

Grade 5 to 12
Format Kit
Product Type Optical Bench
Languages English
Subject Physics - Optics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.