Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Lens Coating | UV-treated |
| Lens Tint | Clear |
| Frame Color | Clear with full foam padded frame |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |