Learn More
Invertase from baker's yeast (S.cerevisiae), 200-300 units/mg, MilliporeSigma™ Supelco™
Catalog No. 111015768 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
11-101-5768 100 mg
1 options
Catalog No. 11-101-5768 Supplier MilliporeSigma Supelco Supplier No. I9274100MG
Add to Cart
Add to Cart
Invertase is encoded by the SUC2 (Saccharomyces cerevisiae S288C) gene. In yeast this enzyme exist in: glycosylated (secreted) and non-glycosylated (intracellular) form. The external invertase is composed of carbohydrate (50%) and 60,000 D subunits.
Food and Beverages and General Analytical
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9001-57-4
Specifications
Packaging Poly Bottle
Quantity 100 mg