Learn More
Johannes Lieder Invertebrata Microscope Slide, Elementary Set
SCI_ED
Catalog No. S139707 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S139707 Cross and Longitudinal Section
1 options
This item is not returnable. View return policy
Catalog No. S139707 Supplier Johannes Lieder Supplier No. 2100
Add to Cart
Add to Cart
This item is not returnable. View return policy
An elementary set of 25 invertebrata microscope slides.
  • 25 Microscope Slides
  • The most important representatives of Protozoa, Sponges, Coelenterata, Vermes, Arthropoda, Insecta Mollusca, Echinodermata, and Acrania.
    • Amoeba proteus w.m.
    • Euglena a common flagellate with eye spot
    • Paramaecium a common ciliate
    • Sycon marine sponge t.s. of body
    • Hydra extended specimen w.m.
    • Dicrocoelium lanceolatum sheep liver fluke w.m.
    • Planaria t.s. of body
    • Taenia saginata tapeworm proglottids in different stages t.s.
    • Trichinella spiralis l.s. of muscle with encysted larvae
    • Lumbricus earthworm t.s. of body in region of typhlosole
    • Daphnia water flea w.m.
    • Cyclops copepod w.m.
    • Spider leg with comb w.m.
    • Spider spinneret w.m.
    • Musca domestica house fly head and mouth parts w.m.
    • Periplaneta cockroach biting mouth parts w.m.
    • Apis mellifica honey bee mouth parts of worker w.m.
    • Musca domestica house fly leg with pulvilli w.m.
    • Apis mellifica wings w.m.
    • Trachea from insect w.m. 21
Specifications
Classification Zoology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Slide Type Prepared Slide