Learn More
Lab-Aids™ Investigating Plant Macronutrients Using Hydroponics
SCI_ED
Catalog No. S20961
Encompass_Preferred
Change view
Click to view available options
Kit: Investigating Plant Macronutrients Using Hydroponics
Catalog No.
S20961 Kit: Investigating Plant Macronutrients Using Hydroponics
1 options
Catalog No. S20961 Supplier Lab-Aids™ Supplier No. 36R
Includes
Teacher's guide; MSDS packet; 28 student worksheets and guides; 36 germinating plugs; 6 greenhouses with detachable vented roofs; 6 germinating floats; 6 light shields for greenhouses; 5 seed packets; 1 bottle of each solution: Iron Chelate, Trace Element, Ca(NO3)2, CaCl2, KCl, KNO3, MgSO4 and KH2PO4
Requires
This lesson requires several weeks of observation following the initial setup lesson
Observe plants growing in a nutrient solution, study and compare seed germination time and plant growth time, observe development of roots, stems and leaves, study effects of nutrient deficiency, use controls in experiments and understand hydroponics.
Do plants only grow in soil? Not always. Sometimes they grow in water. This stimulating and fun kit includes uniquely designed Lab-Aids™ transparent plastic culture chambers that allow students to 'clearly' view plant growth and development in various nutrient solutions. The following experiments are included: Seed germination time comparisons; Plant growth time comparisons; Effects of nutrient deficiency; An understanding of hydroponics. This highly graphic and unique activity can be used individually or in groups at many grade levels. It is an excellent activity for science fairs or projects. Accommodates 2 classes, each with 6 groups of 4 students. Includes Teacher guide, student sheets, assessment questions and SDS for all chemicals.
Specifications
Product Type Kit: Investigating plant nutrients using hydroponics
Includes Teacher's guide; MSDS packet; 28 student worksheets and guides; 36 germinating plugs; 6 greenhouses with detachable vented roofs; 6 germinating floats; 6 light shields for greenhouses; 5 seed packets; 1 bottle of each solution: Iron Chelate, Trace Element, Ca(NO3)2, CaCl2, KCl, KNO3, MgSO4 and KH2PO4
Class Size 24 Students