Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperFi™ Buffers

Catalog No. 12355005
Encompass_Preferred
Click to view available options
Color:
Colorless
Green

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.

Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
For Use With (Application) Hot-start PCR
Quantity 4 x 1.25 mL
Includes 5X SuperFi Buffer (4 x 1.25 mL)

Store at –5°C to –30°C.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.