Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AccuPrime™ Pfx SuperMix

Catalog No. 12344040
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 rxns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-344-040 200 rxns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-344-040 Supplier Invitrogen™ Supplier No. 12344040
  • $852.00
    $809.65 / Each of 1

    Save $42.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

AccuPrime Pfx SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 200 amplification reactions of 25μL each (4 x 1.125mL) are provided. AccuPrime Pfx SuperMix Components: 22U/mL Thermococcus species KOD thermostable polymerase complexed with anti-KOD antibodies, 66mM Tris-SO4 (pH 8.4), 30.8mM (NH4)2SO4, 11mM KCl, 1.1mM MgSO4, 330μM dNTPs, AccuPrime proteins, stabilizers.

Requires

Template and primers

AccuPrime Pfx SuperMix is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.

Invitrogen AccuPrime Pfx SuperMix provides reagents for high-fidelity, high-specificity amplification of DNA fragments. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading activity. This ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs is designed for efficient PCR amplification.

Features

  • >26-times higher fidelity than with Taq DNA polymerase
  • Reduced set-up time
  • Produces products with blunt ends

Applications

  • High-fidelity PCR
  • Amplification of fragments up to 15 kb
  • Cloning & mutagenesis

Notes

  • Thermostable AccuPrime Pfx SuperMix proteins enhance primer-template hybridization during every cycle, increasing specificity and yield.

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.1X
Content And Storage AccuPrime Pfx SuperMix (4 × 1.125 mL)

Store at –20°C in a non-frost-free freezer.
Detection Method Primer-probe
GC-Rich PCR Performance Low
Polymerase AccuPrime Pfx DNA Polymerase
Reaction Speed Standard
Quantity 200 rxns
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 26X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 15 kb or less
Show More Show Less
My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?

Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.

The primers I am using worked for PCR initially, but over time, have stopped working. What happened?

Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.

I don't see a pellet in my oligo tube order. Should I ask for a replacement?

The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.

There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?

If the oligo was overheated, it will appear as a “ball”-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.

There is a green color in my lyophilized oligo. Can I still use it?

If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.

I just received my primers and they look yellow. Can I still use them?

Most of the time the color should not affect PCR or any other experimental application since typically it is caused by the iodine used in the synthesis. There are some exceptions, however. Brown oligos can also be caused by the primer being overdried, and if this is the case, the primer may not work.

My primer has an extra inserted base. How could this happen?

If detritylation occurs inappropriately and/or if the synthesizer has an error and delivers the wrong base, an extra inserted base can occur in your primer. Please contact techsupport@thermofisher.com for assistance.

I'm missing a nucleotide in the middle of my sequence. How could this happen?

There are two possibilities that could occur in any round of extension when creating your primer:

1.The added base is not detritylated correctly, missing one base addition but allowing possible extension in the next round.
2.The trityl group was removed, but not coupled or capped correctly before addition of the next base, allowing the chain to continue.

I ordered a primer with restriction enzyme sites flanking the 3' and 5' ends of my oligo with desalted purification. When trying to subclone the PCR product, I get very few colonies. I have tested all conditions, and it seems to be the oligo causing the problem. Can you explain why this happened?

Better purification of the oligos is recommended to provide you with full-length oligo sequence. Adding restriction sites adds on 10 or more bases to the basic 20-25-mer, making primers longer than 30 bases with a relatively low percentage of full-length sequences after desalting. Additionally, failure sequences occur at the 5' end of the sequence as oligos are generated from 3' to 5' end. Therefore, restriction sites introduced at the 5' end of primers can be compromised, resulting in missing bases.

I'm getting low yield of my oligo upon reconstitution. What happened?

The oligo may not have been fully solubilized. After addition of TE buffer, make sure the oligo was vortexed for a full 30 seconds and/or pipette up and down more than 10 times. Primers may be present along the sides of the tubs, so when resuspending the oligo, the sides of the tubes should be “rinsed” too.

I received my primer order, but the yield is lower than the scale that I ordered. Why is this?

The scale that is ordered refers to the starting synthesis scale, or amount of starting material used to create your oligo. Based on purification and efficiency, you will receive less than the starting synthesis scale. However, we do have a minimum yield guarantee based on the starting synthesis scale which can be found here: https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos/oligo-ordering-details/oligo-minimum-yield-guarantee.html.

I'm seeing high molecular weight EtBr stainable material left in wells. Why is this happening?

This artifact occurs when either too many cycles were performed or too much DNA is added to the reaction. Try heating to 65 degrees C and putting sample on ice before loading.

I'm getting an unexpected product when performing PCR. What could be the cause of this and what do you suggest I try?

Please see the following possibilities and suggestions we have:

-Primer design: try longer primers to avoid binding at alternative sites, avoid 3 consecutive G or C nucleotides at the 3' end.
-Annealing temperature: increase annealing temperature to increase specificity.
-Mg2+ concentration: try a lower concentration.
-DNA contamination: use aerosol tips and separate work area to avoid contamination, use UNG/UDG technique to prevent carryover.

I'm getting no bands from my PCR product. What could cause this?

Here are some reasons why your PCR experiment may be failing:

-NaCl at 50 mM will inhibit the enzyme.
-Too much KCl in the reaction. Do not exceed 50 mM.
-Incorrect annealing temperature was used.
-Incomplete denaturation (time and temperature must be long and high enough).
-Template had long runs of GC's [Woodford et al. (1995) Nucleic Acids Res 23:539 show that by eliminating all potassium from the amplification reactions, GC-rich regions in templates are sufficiently destabilized to allow PCR].
-10% DMSO partially inhibits Taq.
-Hemin (in blood samples) inhibits Taq.
-Use of super-irradiated (treated with >2500 mJ/cm2) mineral oil will either inhibit or decrease yield of PCR product [Dohner (1995) Biotechniques 18:964].
-Do not use a wooden toothpick to pick colonies or scoop out DNA from a gel prior to PCR. It has been reported that this technique can inhibit PCR [Lee (1995) BioTechniques 18:225].
-Other inhibitors of Taq DNA polymerase were present (e.g., indigo dyes, heme). Add BSA to the PCR, increase the amount of Taq, and/or increase the volume of the PCR to dilute out them inhibitor.

I'm getting low yield of my desired fragment. What am I doing wrong and how can I increase my yield?

Please see our suggestions below to increase yield:

-Do not use a wooden toothpick to pick colonies or scoop out DNA from a gel prior to PCR. It has been reported that this technique can inhibit PCR. [Lee (1995) BioTechniques 18:225].
-Not enough enzyme was used.
-Denaturation/extension temperature was too high and enzyme died prematurely.
-Too much DMSO (>10%).
-Incorrect annealing temperature: run a series of reactions using different annealing temperatures, starting 5 degrees below the calculated Tm.
-Too few cycles.
-Insufficient or too much Mg2+.
-Poorly designed primers: double check primer sequence against template sequence, primers should have similar melting temperatures, avoid complementary sequences at the 3' end of primers.
-Carryover inhibitors (e.g., blood, serum).
-Denaturation time was too short. Genomic and viral DNA can require denaturation times of 10 minutes.
-Not a long enough extension time was used depending on the size of product being amplified.
-Use of super-irradiated (treated with >2500 mj/cm2) mineral oil will either inhibit or decrease yield of PCR product [Dohner (1995) Biotechniques 18:964].
-Template had long runs of GC's [Woodford et al. (1995) Nucleic Acids Res 23:539 show that by eliminating all potassium from the amplification reactions, GC-rich regions in templates are sufficiently destabilized to allow PCR]. Alternatively, a combination of 1.0 M betaine with 6-8% DMSO or 5% DMSO with 1.2-1.8 M betaine can be used to amplify GC-rich templates [Baskaran (1996) Genome Res 6:633].
-Other inhibitors of Taq DNA polymerase were present (e.g., indigo dyes, heme, melanin, etc.). Add BSA to the PCR (~160-600 µg/mL), increase the amount of Taq, and/or increase the volume of the PCR to dilute out the inhibitor. The concentration of BSA to add may be dependent on the amount and type of inhibitor present. Additionally, fatty acid-free, alcohol-precipitated BSA, or Fraction V BSA all should be effective.

I'm seeing smearing after PCR. What is causing this?

Please see some reasons below for seeing smearing:

-The enzyme, primer, Mg2+, and/or dNTP concentration was too high.
-The annealing temperature was too low for the primers being used.
-Too many cycles were used.
-The annealing and extension times were too long.
-Bad or old primers.
-Too much template was used initially, try to start with 104-106 molecules
-Consider using additives or PCR Optimizer Kit (Cat. No. K122001), especially if you feel strongly that the primers should work/have worked before and are using Taq.

If I choose mixed bases, e.g., GC, for my oligo manufacturing, will it be a 50/50 mix?

No, we do not guarantee 50/50 of mixed bases. If a mix of GC bases is requested, for example, the synthesizer would deliver half the normal amount of G and half the normal amount of C. Coupling efficiency is not taken into account. Therefore, it is possible that a mix, such as 30/70, will be delivered.

How are these oligos quality controlled?

For 25, 50, and 200 nmol desalted and cartridge-purified DNA oligos, there is 100% A260 analysis. Random samples of 25% of the oligos produced are tested by either capillary electrophoresis or mass spectrometry. DNA oligos that are desalted and ordered at 25 and 50 nmol scales also have 100% real-time digital trityl monitoring during analysis. Desalted DNA oligos ordered at 1 and 10 µmols, DNA oligos at any scale that are purified by HPLC and PAGE, the majority of the DNA oligos with 3' and/or 5' modifications, and RNA oligos have 100% A260 analysis and capillary electrophoresis or mass spectrometry.

How many oligos do I need to order for a 96-well plate order or a 384-well plate order?

The plate orders must contain an average of 24 or more oligos per plate for 96-well plates or 192 or more oligos per plate for 384-well plates across the entire order.

Why do different programs calculate different Tm values?

Tm values are not absolute - they are an approximation of the melting temperature range which exists. A thermal profile for a given oligo shows a 10-15 degree range of melting depending on the amount of salt but also on the base composition and concentration of primer in the reaction which are not precisely defined. One should not rely solely on the given Tm value as the only one that will work. Tm is the temperature at which 50% of the primer and its complementary sequence are present in a duplex DNA molecule. The Tm is necessary to establish an annealing temperature for PCR. Reasonable annealing temperatures range from 55 degrees C to 70 degrees C. Annealing temperatures are generally about 5 degrees C below the Tm of the primers. Since most formulas provide an estimated Tm value, the annealing temperature is only a starting point. Specificity for PCR can be increased by analyzing several reactions with increasingly higher annealing temperatures.

Why are HPLC or cartridge purification not offered for larger oligos?

As oligos increase in length, the column purification is less effective in separating the failure oligos from the correct products. PAGE purification would be the method of choice in this case.

What are Value Oligos?

Value Oligos are the most cost-effective and fastest way to order oligos. They are available for 5-40-mers, at a 25 or 50 nanomole scale, with a range of purification options to suit your needs, and are eligible for next-day delivery. The cost is calculated per oligo as opposed to per base. Value Oligos are not available with modifications. Value Oligos undergo the same QC standards as our standard oligos with the same manufacturing process.

How do I calculate the melting temperature of my primers?

A common equation used to calculate primer Tm is as follows: Tm (in degrees C) = 2 (A+ T) + 4 (G + C)

What type of modifications does Life Technologies offer for my primers?

Please take a look at this list (https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos/oligo-ordering-details/oligo-modification-options.html) of standard modification options that we offer. If you do not see the modification option you would like, please email our Technical Support team at techsupport@thermofisher.com to see if we can accommodate your request.

How do I determine the percentage of full-length oligonucleotide?

The percentage of full-length oligonucleotide depends on the coupling efficiency of the chemical synthesis. The average efficiency is close to 99%. To calculate the percentage of full-length oligonucleotide, use the formula: 0.99n-1. Therefore, 79% of the oligonucleotide molecules in the tube are 25-bases long; the rest are <25 bases. If you are concerned about starting with a preparation of oligonucleotide that is full-length you may want to consider cartridge, PAGE, or HPLC purification.

Why is coupling efficiency important?

Coupling efficiency is important as the effects are cumulative during DNA synthesis. The numbers below shows the effect of a 1% difference in coupling efficiency and how this influences the amount of full-length product available following synthesis of different length oligos. Even with a relatively short oligo of 20 bases, a 1% difference in coupling efficiency can mean 15% more of the DNA present following synthesis is full-length product.

Number of bases added, 99% coupling full-length, Failures, 98% coupling full-length, Failures:
- 1, 99, 1, 98, 2
- 2, 98.01, 1.99, 96.04, 2.96
- 3,97.03, 2.97, 94.12, 5.88
- 10, 90.44, 9.56, 81.71, 18.29
- 20, 81.79, 18.21, 66.76, 33.24
- 30, 73.79, 26.03, 54.55, 63.58
- 50, 60.5, 39.5, 36.42, 63.58
- 95, 38.49, 61.51, 14.67, 85.33

What are the minimum yield guarantees you offer for your oligos?

The scale of synthesis is the starting point for synthesis, not the guaranteed final amount. We guarantee the total yield of oligonucleotide as a minimum number of OD units. Use this link (https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos/oligo-ordering-details/oligo-minimum-yield-guarantee.html) for the minimum yield guarantees we offer for our oligos.

Do you have any resources to help design primers?

Yes. OligoPerfect Designer can be used to design primers for sequencing, cloning, or detection.

Can you suggest some guidelines that will help me design my PCR primers?

These guidelines may be useful as you design your PCR primers:

- In general, a length of 18-30 nucleotides for primers is good.
- Try to make the melting temperature (Tm) of the primers between 65 degrees C and 75 degrees C, and within 5 degrees C of each other.
- If the Tm of your primer is very low, try to find a sequence with more GC content, or extend the length of the primer a little.
- Aim for the GC content to be between 40 and 60%, with the 3' of a primer ending in C or G to promote binding.
- Typically, 3 to 4 nucleotides are added 5' of the restriction enzyme site in the primer to allow for efficient cutting.
- Try to avoid regions of secondary structure, and have a balanced distribution of GC-rich and AT-rich domains.
- Try to avoid runs of 4 or more of one base, or dinucleotide repeats (for example, ACCCC or ATATATAT).
- Avoid intra-primer homology (more than 3 bases that complement within the primer) or inter-primer homology (forward and reverse primers having complementary sequences). These circumstances can lead to self-dimers or primer-dimers instead of annealing to the desired DNA sequences.
- If you are using the primers for cloning, we recommend cartridge purification as a minimum level of purification.
- If you are using the primers for mutagenesis, try to have the mismatched bases towards the middle of the primer.
- If you are using the primers for a PCR reaction to be used in TOPO cloning, the primers should not have a phosphate modification.
Read more about primer design tips and tools at https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos/primer-design-tools.html.

How does a two-temperature protocol work and when would you suggest using one?

You may choose to do a two-temperature protocol when the annealing temperature is relatively high. In this case, you would combine the annealing and the elongation steps, i.e., both can occur together at a temperature >62 degrees C. The advantage of a two-temperature protocol is that it is considerably quicker in comparison to the conventional three-temperature protocol.

How can I facilitate the amplification of templates with hairpin-loop structures or high GC-content?

You can try adding 5-10% DMSO, up to 10% glycerol, or 1-2% formamide or a combination of these to facilitate difficult templates. Note: the use of cosolvents will lower the optimal annealing temperatures of your primers.

Why is it difficult to amplify a GC-rich template?

A GC-rich template often has a higher melting temperature and may not denature completely under the normal reaction conditions.

What does hot start PCR mean?

Hot start is a way to prevent DNA amplification from occurring before you want it to. One way to do this is to set up the PCR reaction on ice, which prevents the DNA polymerase from being active. An easier method is a use a ‘hot-start' enzyme, in which the DNA polymerase is provided in an inactive state until it undergoes a high-heat step.

What are the main steps in PCR?

The main steps are: denaturation, annealing, and extension. The template is typically heated to a high temperature (around 94-95 degrees C) allowing for the double-stranded DNA to denature into single strands. Next, the temperature is lowered to 50-65 degrees C, allowing primers to anneal to their complementary base-pair regions. The temperature is then increased to 72 degrees C, allowing for the polymerase to bind and synthesize a new strand of DNA.

What is the difference between Platinum technology and AccuPrime technology?

With Platinum technology, anti-DNA polymerase antibodies bind to the enzyme until the denaturing step at 94 degrees C, when the antibodies degrade. The polymerase is now active and primer extension can occur. AccuPrime Taq combines Platinum Taq (Taq + Platinum antibodies) with proprietary thermostable AccuPrime accessory proteins. The 10X reaction buffer contains the accessory proteins which enhance specific primer-template hybridization during each cycle of PCR.

What PCR enzyme would you recommend for use with the Directional TOPO Cloning Kits?

For the Directional TOPO Cloning Vectors, a PCR product must be generated by a proofreading enzyme to create a blunt product. Pfx50 or Accuprime Pfx and Accuprime Pfx Supermix from Thermo Fisher Scientific are recommended for use.

When cloning a Pfx-amplified PCR product, the insert to vector ratio is an important consideration. The PCR product generally needs to be diluted since Pfx generates a high concentration of product and using too much insert DNA can hamper the TOPO reaction. A 1:1 molar ratio of vector to insert (or about 2-10ng of insert) is recommended.

Do Elongase and Platinum Taq High Fidelity enzymes leave a 3'-A overhang on the PCR product for subsequent cloning into a TOPO TA Cloning or original TA vectors? What about Platinum Pfx polymerase?

Elongase and Platinum Taq High Fidelity polymerase mixes do leave 3' A overhangs on a portion of the PCR products, however, the cloning efficiency is greatly reduced from that obtained with Taq polymerase alone. Platinum Pfx polymerase does not leave 3' A overhangs. Therefore, with all proofreading enzymes or enzyme mixes that contain proofreading polymerases, we recommend that you treat the PCR product with Taq at the end of the PCR reaction, prior to TA cloning. To do this, add 1 U of Taq to a 50 µL reaction and incubate at 68-72 degrees C for 15 min. Phenol extract and ethanol precipitate the product before TA cloning.

Additional notes: The cloning efficiency decreases with increasing size of PCR products. For larger PCR fragments, we recommend that you gel-purify the PCR product and screen several clones. PCR primers should be designed with a 5' G, since Taq leaves a 3' A overhang preferentially on DNA ending in C.
Reference: Hu (1993) DNA and Cell Biology 12:763.
TA Cloning reference: Mead, D.A., Pey, N.K., Herrnstadt, C., Marcil, R.A., and Smith, L.M. (1991) BioTechnology 9, 657.

How should I adjust the Platinum Pfx DNA Polymerase protocol if I am trying to generate an amplicon greater than 2 kb, or if I am starting with long PCR primers?

To generate amplicons greater than 2 kb, use 2.5 units of Platinum Pfx polymerase (instead of 1 unit), decrease the extension temperature to 68 degrees C, and increase the extension time to 1 kb/minute.

If long PCR primers are used with Platinum Pfx polymerase, increase the magnesium concentration in the reaction to 1.5 mM.

Why did I get a lower yield of long PCR product with Platinum Pfx than with a polymerase mix such as Elongase enzyme?

While enzyme mixes offer improved fidelity over Taq DNA Polymerase alone, Platinum Pfx DNA Polymerase provides much higher fidelity since it is a proofreading polymerase exclusively. If yield is more important than fidelity, then an enzyme mix such as Platinum Taq High Fidelity or Elongase enzyme mix is a better choice of enzyme (since they contain a significant amount of Taq in addition to the proofreading polymerase). Platinum Pfx does give high yields relative to other proofreaders, but may not give as great a yield as the enzyme mixes.

Why doesn't Pfx DNA Polymerase yield the same quantitiy of PCR fragment as other thermostable proofreading polymerases?

Generally, it should. However, the 10X Pfx Amplification buffer is optimized to work with Platinum Pfx DNA Polymerase, so it is important Pfx is not used with buffers meant for other polymerases. Additionally, since the magnesium concentration is lower in the Pfx buffer, the primer annealing temperature may need to be lowered.

What is the fidelity of Platinum Pfx DNA polymerase (please quantitate)?

The rpsL fidelity assay was used to generate this data. Briefly, a plasmid containing an AmpR and SmS gene was amplified by PCR and religated. Transformations were plated on Amp plates and Amp/Sm plates. The mutant frequency = rpsL mutant colonies/total colonies x 100.
Results from this assay:
Taq: 4.8/100,000
Platinum Pfx: 1/1,000,000
Pfu were 1.5/1,000,000


WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.