Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anchored Oligo(dT)20 Primer

Catalog No. 12577011
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-577-011 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-577-011 Supplier Invitrogen™ Supplier No. 12577011
  • $234.00
    $215.65 / Each of 1

    Save $18.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC).

Anchored Oligo(dT)20 Primer is a primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (dG, dA, or dC) and then by dN (dA, dT, dG, or dC). Due to the variable end-sequence, the primer is anchored to the 5'-end of the poly(A) tail of mRNA and prevents priming within the poly(A) tail.

Anchored Oligo(dT)20 Primer is recommended for use in cDNA labeling protocols and in RT-PCR. It is commonly used for first-strand cDNA systhesis.

Quality control: This product is qualified in a first-strand cDNA synthesis reaction

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage • Anchored Oligo(dT)20 Primer in DEPC-treated water (20 μL at 2.5 μg/μL)

Store at –20°C.
Guaranteed stable for 6 months when properly stored.
5'Primer Modification Phosphate
Primer Length 22-mer
Primer Sequence 5'd PO4 [(T)20VN]3'
Purification Method Gel-purified, Desalted
Shipping Condition Dry Ice
Concentration 2.5 μg/μL
Primer Anchored Oligo dT Primer
Quantity 20 μL
For Use With (Application) First-strand cDNA Synthesis
Show More Show Less
How does the Anchored Oligo(dT)20 Primer differ from standard oligo(dT) primers?

Anchored Oligo(dT) primers have 2 random bases at the 3' end of the stretch of Ts. The first random base is either an A, G, or C, while the second can be any of the 4 standard nucleotides, i.e., A, T, G or C. The use of anchored oligo(dT) primers results in increased cDNA synthesis yields.

For first-strand cDNA synthesis, is it better to use oligo(dT), random hexamers, gene-specific primer (GSP), or combination of these primers?

The choice of primer depends on your experimental goals. Oligo(dT) is recommended when using total RNA for cDNA synthesis. It is the key to full-length cDNA synthesis. Random hexamers give a series of short first-strand products spanning the entire mRNA. Use of random hexamers may be helpful if the PCR fragment is at the 5´ end of a large mRNA. To ensure full-length cDNA synthesis of large transcripts, oligo(dT) can be added along with random hexamers during first-strand synthesis. Gene-specific primers (GSP) for cDNA synthesis may also be used and are required in a few applications such as 5´ RACE and qRT-PCR. For GC-rich templates or templates rich in secondary structure, a GSP may not work as well as priming with oligo dT for first-strand synthesis. If an RT-PCR is problematic, trying different options of oligo dT, random primers and/or GSP for priming first-strand synthesis may resolve the issue. Oligo(dT)20 primer (Cat. No. 18418-020) is recommended for use with SuperScript III Reverse Transcriptase (Cat. no. 18080-044), ThermoScript Reverse Transcriptase (Cat. No. 12236-014), Thermo-X Reverse Trascriptase (Cat. No. 11150-025), and Cloned AMV Reverse Transcriptase (Cat. No. 12328-019).

Reference:
Frohman,M.A., Dush,M.K., Martin, G.R. (1988) Proc. Nat. Acad. Sci USA 85, 8998

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.