Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biotin-14-dATP

Catalog No. 19524016
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-524-016 50 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-524-016 Supplier Invitrogen™ Supplier No. 19524016
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Biotin-14-dATP is supplied at 0.4mM in 100mM Tris-HCl (pH 7.5), 0.1mM EDTA.

A patented dATP analog with biotin attached at the 6-position of the purine base by a 14-atom linker

The long, 14-atom spacer arm allows accessibility to streptavidin for sensitive detection of probe-target hybrids in in situ and filter-based hybridizations. Biotin-14-dATP can be efficiently incorporated into DNA by nick translation in the presence of dCTP, dGTP, and dTTP. Biotin-14-dATP can be added to the 3' end of DNA using terminal deoxynucleotidyl transferase. The amount supplied is sufficient for labeling up to 50μg of DNA by nick translation.

Performance and Quality Testing: Purity is evaluated by reverse-phase HPLC analysis.

DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.4 mM
Content And Storage Biotin-14-dATP is supplied at 0.4 mM in 100 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.
Format Liquid
Labeling Method Direct Labeling
Label or Dye Biotin and Other Hapten
Product Type Labeled Nucleotide
Quantity 50 nmol
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.