Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Serum Albumin

Catalog No. 15561020
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-561-020 150 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-561-020 Supplier Invitrogen™ Supplier No. 15561020
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use as enzyme stabilizer

Bovine Serum Albumin is suitable for use as enzyme stabilizer during purification or for dilution of restriction endonucleases and nucleic acid modifying enzymes. BSA is also commonly used in DNA and protein labeling experiments as a blocking agent to minimize background.

  • Acetylated to inactivate contaminating nucleases and proteases
  • Supplied as aqueous solution at concentration of 45 to 55mg/mL
  • Performance and quality testin: Purity is determined by SDS-PAGE
  • No detectable contaminating activity is observed in 3' and 5' exonuclease, DNA nicking, and restriction endonuclease inhibition assays

ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 45 to 55 mg/mL
Content And Storage Bovine Serum Albumin is supplied as an aqueous solution at a concentration of 45 to 55 mg/ml. Store at -20°C. Bovine Serum Albumin is guaranteed for 6 months from date of purchase unless otherwise specified in product literature.
Protein BSA
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 150 mg
Product Type Bovine Serum Albumin
What is the Bovine Serum Albumin (BSA) (Cat. No. 15561020) dissolved in?

BSA is supplied sterile in deionized water.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.