Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Streptavidin Magnetic Beads

Catalog No. 65607D
Encompass_Preferred
Change view
Click to view available options
Product Type:
Dynabeads™ M-270 Streptavidin
Dynabeads™ M-280 Streptavidin
Dynabeads™ MyOne™ Streptavidin C1
Dynabeads™ MyOne™ Streptavidin T1
Dynabeads™ Streptavidin Trial Kit
Dynabeads™ Streptavidin for Target Enrichment
Dynabeads™ kilobaseBINDER™ Kit
Quantity:
10 mL
100 mL
2 mL
200 Isolations
4 x 1 mL
50 mL
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
65607D Dynabeads™ Streptavidin for Target Enrichment 50 mL
65-001 Dynabeads™ MyOne™ Streptavidin C1 2 mL
65-002 Dynabeads™ MyOne™ Streptavidin C1 10 mL
65-604-D Dynabeads™ MyOne™ Streptavidin T1 50 mL
65-602 Dynabeads™ MyOne™ Streptavidin T1 10 mL
65-601 Dynabeads™ MyOne™ Streptavidin T1 2 mL
65605D Dynabeads™ Streptavidin for Target Enrichment 2 mL
65606D Dynabeads™ Streptavidin for Target Enrichment 10 mL
11-205-D Dynabeads™ M-280 Streptavidin 2 mL
11-206-D Dynabeads™ M-280 Streptavidin 10 mL
60-210 Dynabeads™ M-280 Streptavidin 100 mL
65-305 Dynabeads™ M-270 Streptavidin 2 mL
65-306 Dynabeads™ M-270 Streptavidin 10 mL
60-101 Dynabeads™ kilobaseBINDER™ Kit 200 Isolations
65-801-D Dynabeads™ Streptavidin Trial Kit 4 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 65607D Supplier Invitrogen™ Supplier No. 65607D
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Includes

Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% NaN3 added as preservatives.

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are compatible with high-throughput automation platforms, including KingFisher sample purification systems, ensuring seamless integration into new and existing workflows.

Key advantages of Dynabeads Streptavidin Magnetic Beads

  • ISO 13485–certified manufacturing—exceptional manufacturing control to ensure robust and consistent performance between production lots, meeting international quality standards for medical devices
  • Monosized, monodispersed magnetic beads—uniform particle size provides consistent and reproducible results across applications
  • Dual size options—available in 1-μm and 2.8-μm diameter specifications to accommodate diverse experimental requirements and application-specific needs
  • Multiple surface chemistry options—various surface functionalization chemistries available to optimize binding efficiency and compatibility for specific applications
  • Application-optimized design—size and surface chemistry combinations enable best-fit selection for targeted experimental protocols and assay requirements

Precision-engineered manufacturing delivers exceptional control over critical bead properties

Dynabeads Streptavidin products are based on magnetic polystyrene-based beads that are covalently coupled with recombinant streptavidin, creating a robust platform for target capture and isolation using biotinylated affinity binders. Dynabeads Streptavidin product manufacturing enables exceptional precision over three critical parameters: size distribution, magnetization properties, and surface chemical characteristics. This ensures consistent performance across batches and applications, providing the reliability necessary for reproducible experimental outcomes.

Optimized size selection enhances application-specific performance

The Dynabeads Streptavidin product line offers two distinct size options, each engineered to excel in specific experimental contexts. The 1-μm beads, available in the Dynabeads MyOne series, are ideally suited for applications requiring fast reaction kinetics, slow sedimentation rates, and high sensitivity-to-capacity ratios. These smaller beads provide enhanced surface area contact and reduced gravitational settling, making them particularly valuable for sensitive detection assays.

In contrast, the 2.8-μm beads, found in the Dynabeads M-270 and M-280 series, are perfect for applications demanding low background binding and reduced risk of aggregation in aggregation-prone applications. These larger beads offer even easier handling and faster magnetic separation compared to the Dynabeads MyOne series, facilitating streamlined workflow processing.

Tailored surface chemistry options address diverse experimental requirements

The Dynabeads Streptavidin product line includes two distinct surface chemistry configurations designed to optimize performance across varied experimental conditions. Hydrophobic surfaces are available in Dynabeads MyOne Streptavidin T1, Dynabeads Streptavidin for Target Enrichment, and Dynabeads M-280 Streptavidin products. These surfaces are BSA-blocked after streptavidin coupling making them ideal for high signal-to-noise applications such as immunoassays, target enrichment, and on-bead PCR for next-generation sequencing.

Alternatively, hydrophilic surfaces are featured in Dynabeads MyOne Streptavidin C1, Dynabeads Streptavidin for In Vitro Transcription, and Dynabeads M-270 Streptavidin products. These surfaces contain no BSA blocking and are particularly suitable for hydrophobic targets, harsh lysis conditions, sample preparation workflows for mass spectrometry, and applications demanding low or no nuclease activity, such as on-bead reverse transcription or in vitro transcription.

For researchers uncertain about which bead type best suits their application, a Dynabeads Streptavidin Trial Kit is available. It allows for side-by-side performance evaluation of different bead formulations under your specific experimental conditions, ensuring optimal selection for your research needs.

ISO 13485 certification ensures scalable quality and OEM support

Manufactured under ISO 13485 certification, Dynabeads Streptavidin products deliver outstanding batch-to-batch consistency. This robust quality assurance framework ensures that once customers develop an assay or workflow, they can rely on reproducible results across multiple experimental iterations. This consistency makes these versatile beads an efficient solution for applications spanning the entire spectrum from fundamental research to commercial manufacturing processes. For researchers developing commercial products or services, or those requiring larger bulk volumes, specialized support is available by emailing oemdynal@thermofisher.com.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic polystyrene-based beads covalently coupled with recombinant Streptavidin
Concentration 10 mg/mL
Content And Storage Dynabeads Streptavidin for Target Enrichment is supplied in PBS, pH 7.4, with 0.1% BSA, 0.02% NaN3. Store at 2°C to 8°C.

Format Beads in Suspension
Isolation Technology Magnetic Bead
Diameter (Metric) 1 μm (CV < 5%)
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) Capture of targeted DNA sequences for Next-Generation Sequencing (NGS) and hybridization-based DNA assays
High-throughput Compatibility High-throughput Compatible
Product Line Dynabeads
Product Type Dynabeads™ Streptavidin for Target Enrichment
Quantity 50 mL
Shipping Condition Ambient Temperature
Binding Property 950-1500 pmoles free biotin/mg beads
Regulatory Status For Research Use Only
Certifications/Compliance ISO9001 and ISO13485
Surface Functionality Tosylactivated hydrophobic surface blocked with BSA
Show More Show Less
What range of DNA fragment sizes can be captured with Dynabeads Streptavidin for Target Enrichment (Cat. No. 65605D)?

We do not have hard cutoffs for the lengths of the fragments that can be captured with our Dynabeads products, including Cat. No. 65605D. In general terms, however, the beads should work well with any fragment up to 2kb. If you have mixed sizes in the sample, you can expect the smaller fragments being captured more efficiently than the longer ones.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.