Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Opener Replacement Steel Blades

Catalog No. G535010
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 blades
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G535010 10 blades
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G535010 Supplier Invitrogen™ Supplier No. G535010
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A simple device specifically designed to quickly and efficiently open an E-Gel™ cassette

Allows purification of DNA fragments from the gel, transfer samples onto a membrane for Southern blot analysis, or post-stain Clear EGels. The E-Gel Opener is safe and easy to use. Simply place the E-Gel cassette into the E-Gel Opener and turn the knob to tighten.

  • The E-Gel Opener uses two steel blades to safely and quickly pop open the E-Gel cassette without harming gel
  • In only a few minutes and with minimal effort, E-Gel will be ready for subsequent procedures
  • Anodized aluminum construction for maximum durability

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) E-Gel™ Cassettes
Type Gel Opener Replacement Steel Blade
Quantity 10 blades
How can I get better separation of my bands?

First check the percentage of your agarose gel. A higher percentage will help you to resolve smaller molecular weights while a lower percentage will help you to resolve larger molecular weights.

How much DNA can I load for my E-Gel Agarose Gels?

To determine the amount of DNA to load per well for your specific E-Gel agarose gel, please refer to the table on page 14 of the E-Gel Power Snap Electrophoresis System user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017050_egel_powersnapsystem_UG.pdf).

What does intact RNA look like when run on an agarose gel?

Intact RNA should have a 2:1 ratio of 28S:18S bands. You may see a smear of RNA that extends from <9 kb to 0.5 kb, indicating the presence of mRNA in the sample. To see an image or to read more about RNA assessment, visit this website (https://www.thermofisher.com/us/en/home/references/protocols/nucleic-acid-purification-and-analysis/rna-protocol/agarose-gel-electrophoresis-of-rna.html).

How can I perform RNA electrophoresis?

For nondenaturing RNA electrophoresis, we recommend using our E-Gel Precast Agarose Gels. Please note that E-Gel Agarose Gels are not validated to be RNAse-free. However, many of our customers routinely use E-Gel Agarose Gels for RNA analysis with success. If RNA is run on an E-Gel Agarose Gel, any loading buffer that would be used for nondenaturing RNA electrophoresis should be fine.

For denaturing RNA electrophoresis, there are several denaturing agents to choose from, including formaldehyde, glyoxal, formamide, and methyl mercury. Denaturing conditions disrupt hydrogen bonding so that RNA runs without secondary structure, as single-stranded molecules.

For denaturing RNA electrophoresis, our E-Gel EX Agarose Gels can be used. The only denaturing agent that is compatible with the E-Gel EX system is formamide, 50-90%. Using other denaturing agents will result in poor band separation and morphology. Please note that we do not recommend running samples prepared in RNA loading buffer on the same gel with samples prepared in water. Please see below for the RNA loading buffer recipe and denaturing electrophoresis conditions:

RNA Loading Buffer:
Deionized formamide: 200 µL
10X MOPS-EDTA-Sodium Acetate Buffer (0.4 M MOPS, pH 7.0, 0.1 M sodium acetate, 10 mM EDTA): 40 µL
Deionized formaldehyde: 76 µL
Water: 14 µL

Denaturing Electrophoresis Conditions:
1. Mix 15 µL of RNA loading buffer with 1-5 µL of RNA (1-5 µg).
2. Heat samples at 65 degrees C for 10 min to denature RNA.
3. Place samples on ice immediately after heating.
4. Load entire sample onto an E-Gel EX agarose gel.
5. Electrophorese for 30 minutes.

For denaturing RNA electrophoresis under formaldehyde-free conditions, we recommend using our NorthernMax-Gly Kit (Cat. No. AM1946). With this kit, RNA samples are denatured in glyoxal/DMSO loading buffer and run on a glyoxal-containing agarose gel.

Should I use agarose or polyacrylamide gels for DNA electrophoresis?

Agarose is commonly used as it is nontoxic, easy to use, and offers a broad range of separation. We offer precast E-Gel Agarose Gels or reagents to pour your own agarose gels. Polyacrylamide gels are typically used for high resolution of DNA molecules that range in size from 10-3,000 bp. We offer precast Invitrogen TBE polyacrylamide gels and UltraPure reagents.

Why do circular and linear DNA migrate differently in gel electrophoresis?

Migration speed through gels is correlated to the compactness of the DNA molecule. Circular DNA can exist as a relaxed circle (if one of the strands is nicked), and this form migrates very slowly. If the DNA circle is supercoiled, it adopts a more compact form and will migrate much more quickly. If both strands of the DNA circle are cleaved and the molecule is linear, that form will also migrate more quickly than the relaxed circle, but not as quickly as the supercoiled circle.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.