Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human miR RNAi Select

Catalog No. 100002318
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-000-2318 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-000-2318 Supplier Invitrogen™ Supplier No. 100002318
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easily design effective RNAi molecules for reliable results

  • Using BLOCK-iT™ RNAi Designer, design Custom BLOCK-iT siRNA sequences for efficient and specific gene expression knockdowns in any organism
  • For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
RNAi Type siRNA
Product Type Human miR RNAi Select
Quantity 1 Each
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.