Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M-MLV Reverse Transcriptase Buffer

Catalog No. 18057018
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-057-018 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-057-018 Supplier Invitrogen™ Supplier No. 18057018
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

1mL M-MLV Reverse Transcriptase Buffer and 500μL DTT (0.1M)

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).

5X buffer composition

  • 250 mM Tris-HCl (pH 8.3)
  • 375 mM KCl
  • 15 mM MgCl2

Included with

  • M-MLV Reverse Transcriptase
  • SuperScript Reverse Transcriptase
  • SuperScript II Reverse Transcriptase
  • SuperScript III Reverse Transcriptase

M-MLV Reverse Transcriptase Buffer does NOT contain actinomycin D, nucleotides, or dithiothreitol (DTT).

Order Info

Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
For Use With (Application) Reverse Transcription
Quantity 1 mL
Includes

• M-MLV Reverse Transcriptase Buffer (1 mL)
• 0.1 M DTT (500 μL)

Store at –20°C.

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

What is the highest temperature that MMLV, SuperScript II, SuperScript III, or SuperScript IV RTs can be used?

The optimal temperature for for MMLV is 42 degrees C. The optimal temperature for SuperScript II RT is 42 degrees C, and can be used up to 50 degrees C. The optimal temperature for SuperScript III RT is 50 degrees C, and can be used up to 55 degrees C. For some qRT-PCR reactions where gene-specific primers are used, you can do the RT reaction at 60 degrees C. The optimal temperature for SuperScript IV RT is 50 degrees C, but can be used up to 65 degrees C.

Can I use elevated temperatures for reverse transcription with Moloney Murine Leukemia Virus (M-MLV) reverse transcriptase?

Moloney Murine Leukemia Virus (M-MLV) Reverse Transcriptase is not well-suited for elevated temperatures. Typically, M-MLV RT operates optimally at temperatures around 37 degrees C to 42 degrees C.

To perform reverse transcription at elevated temperatures (e.g., 50 degrees C or higher), consider using thermostable reverse transcriptases such as SuperScript IV or Maxima H Minus, which are engineered to perform efficiently at higher temperatures.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.