Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M13KO7 Helper Phage

Catalog No. 18311019
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-311-019 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-311-019 Supplier Invitrogen™ Supplier No. 18311019
  • $364.00
    $342.65 / Each of 1

    Save $21.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use with M13/phagemid hosts for the production of ssDNA from vectors containing an f1or f1-like origin of replication

Prepared by modification of procedure of Vieira and Messing, this high-titer helper phage eliminates need to make and maintain a separate stock of helper phage. It contains Kmr gene and p15A origin of replication that allows it to replicate like plasmid inside cells. Detailed protocols for production of ssDNA are provided on product profile.

  • Convenient preparation of ssDNA from M13 or phagemid vectors
  • >1 x 1011 pfu/mL

Cloning, Mutagenesis

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Shipping Condition Dry Ice
Quantity 5 x 1 mL
Product Type M13KO7 Helper Phage
Do you offer an alternative for M13KO7 Helper Phage (Cat. No. 18311019)?

We do not offer an alternative for M13KO7 Helper Phage.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.