Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAX Efficiency™ DH5αF'IQ Competent Cells

Catalog No. 18288019
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-288-019 5 x 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-288-019 Supplier Invitrogen™ Supplier No. 18288019
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

3mL of lawn cells and a vial of monomer M13mp19 RF DNA as a control

MAX Efficiency DH5αF'IQ Competent Cells are high transformation efficiency chemically competent cells prepared by a patented modification of the procedure of Hanahan. This strain is a derivative of the DH5α strain, sharing the same benefits.

MAX Efficiency DH5αF´IQ Competent Cells are high transformation efficiency chemically competent cells prepared by a patented modification of the procedure of Hanahan. This strain is a derivative of the DH5α strain, sharing the same benefits. The DH5αF´IQ strain contains an F´ episome and is a host for M13mp cloning vectors. It also supports cloning plasmids with an f1-like origin of replication (phagemids) or vector that uses expression from the lac promoter. DH5αF´IQ cells can yield >1 x 108 plaque forming units/μg M13mp19 RF with non-saturating amounts (5 pg) of DNA.

MAX Efficiency DH5αF´IQ Competent Cells have the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. They also carry the recA1 mutation which helps to reduce the rate of recombination while propagating plasmid DNA and the endA1 mutation that increases plasmid DNA yield and quantity.

MAX Efficiency DH5αF´IQ Competent Cells offer:
• >3 x 108 transformants/μg efficiency
• >1 × 108 plaque forming units/μg M13mp19 RF
• Stable F´ episome that eliminates need for growth on minimal media
• lacZΔM15 marker that provides α-complementation of the β-galactosidase gene allowing blue/white screening on agar plates containing X-Gal or Bluo-Gal
• Plasmid DNA maintenance with minimum recombination events due recA1 mutation
• endA1 mutation that enables increased plasmid yield and quantity

Note: DH5αF'IQ lawn cells are provided to allow for plaque formation and a vial of monomer M13mp18 RF DNA are supplied as a control.

Genotype
F– φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ–thi–1 gyrA96 relA1/F′ [proAB+lacIqZΔM15 zzf::Tn5(KmR)] 
 
Find the strain and format that fits your needs
• The DH5α strain and its derivatives are available with a variety of transformation efficiencies and in both electrocompetent and chemically competent formats.
• DH5α-T1R competent cells are available in MultiShot format for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome Yes
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • MAX Efficiency DH5αF'IQ Competent Cells (5 x 200 μL)
• F'IQ Lawn Cell Assy (2 x 1.5 mL)
• M13mp18 RF DNA (1 vial)
Store at –80°C.

• S.O.C. Medium (2 x 6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Kanamycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Promoter Laclq
Format Tube
Product Line MAX Efficiency
Quantity 5 x 200 μL
Show More Show Less
How do you recommend that I prepare my DNA for successful electroporation of E. coli?

For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.

The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.

Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.

Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.

You offer competent cells in Subcloning Efficiency, Library Efficiency and MAX Efficiency. How do these differ?

There are a few exceptions, but in general the difference is in guaranteed transformation efficiency as follows:

Subcloning Efficiency cells are guaranteed to produce at least 1.0 x 10E6 transformants per µg of transformed pUC19 or pUC18 supercoiled plasmid
Library Efficiency cells are guaranteed to produce at least 1.0 x 10E8 transformants per µg pUC19 or pUC18 DNA
MAX Efficiency cells are guaranteed to produce at least 1.0 x 10E9 transformants per µg pUC19 or pUC18 DNA

Do any Invitrogen competent cells contain DMSO in the freezing medium?

Yes, several of our competent cells products are frozen with DMSO. The presence of DMSO (dimethylsulfoxide) will generally be indicated in the MSDS files if you have a question about a particular product, but here is a list of commonly used products that are known to have DMSO in the freezing buffer:

One Shot OmniMAX 2 T1 Phage Resistant Cells, Cat. No. C8540-03

One Shot INV?F' Chemically Competent Cells, Cat. No. C2020-03 and C2020-06

One Shot MAX Efficiency DH5?-T1 Chemically Competent Cells, Cat. No. 12297-016

MAX Efficiency DH5?-T1 Phage Resistant Cells, Cat. No. 12034-013

MAX Efficiency DH5? Chemically Competent Cells, Cat. No. 18258-012

Library Efficiency DH5? Chemically Competent Cells, Cat. No. 18263-012

MAX Efficiency DH5? F'IQ Cells, Cat. No. 18288-019

MAX Efficiency Stbl2Chemically Competent Cells, Cat. No. 10268-019

When should DMSO, formamide, glycerol and other cosolvents be used in PCR?

Cosolvents may be used when there is a failure of amplification, either because the template contains stable hairpin-loops or the region of amplification is GC-rich. Keep in mind that all of these cosolvents have the effect of lowering enzyme activity, which will decrease amplification yield. For more information see P Landre et al (1995). The use of co-solvents to enhance amplification by the polymerase chain reaction. In: PCR Strategies, edited by MA Innis, DH Gelfand, JJ Sninsky. Academic Press, San Diego, CA, pp. 3-16.

Additionally, when amplifying very long PCR fragments (greater than 5 kb) the use of cosolvents is often recommended to help compensate for the increased melting temperature of these fragments.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.