Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ One Shot™ MAX Efficiency™ DH10B T1 Phage-Resistant Cells

Catalog No. 12331013
Encompass_Preferred
Change view
Click to view available options
Quantity:
21 x 50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-331-013 21 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-331-013 Supplier Invitrogen™ Supplier No. 12331013
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

One Shot MAX Efficiency DH10B T1 Phage-Resistant E. coli have all the benefits of the DH10B genotype, with the addition of the tonA (also known as fhuA) genotype for resistance to T1 and T5 phage infection.

One Shot MAX Efficiency DH10B T1 Phage-Resistant E. coli have all the benefits of the DH10B genotype, with the addition of the tonA (also known as fhuA) genotype for resistance to T1 and T5 phage infection. The inclusion of the tonA genotype offers added security to protect valuable clones and libraries. MAX Efficiency DH10B T1 cells can achieve high transformation efficiency of >1 x 109 cfu/μg supercoiled plasmid, making them ideal for generating cDNA or genomic libraries, transforming large plasmids (>150 kb in size), and for situations when DNA is limiting.

MAX Efficiency DH10B T1 Phage-Resistant Cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr), allowing construction of genomic libraries of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. This strain also has the lacZΔM15 genotype, providing the option of blue-white screening on plates containing either X-Gal or Bluo-Gal. Finally, the endA1 mutation makes DH10B-T1R cells an excellent vehicle for amplifying plasmid DNA for subsequent extraction and purification.

MAX Efficiency DH10B T1 Phage-Resistant Cells offer:
• Transformation efficiencies of >1 x 109 cfu/μg
• tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• lacZΔM15 marker that provides α-complementation of the β-galactosidase gene allowing blue/white screening on agar plates containing X-Gal or Bluo-Gal
• mcrA genotypic marker and the mcrBC, mrr deletion that allows for cloning DNA that contains methylcytosine and methyladenine
• endA1 mutation that enables increased plasmid yield and quantity

Convenient and efficient format 
DH10B-T1R E. coli are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation so there are no efficiency-zapping, freeze-thaw cycles, or money wasted on unused cells.

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara, leu)7697 galU galK λ- rpsL nupG tonA 
 
Find the strain and format that fit your needs
• We offer other DH strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• The DH10B-T1R strain is available in Multishot format for high throughput applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot MAX Efficiency DH10B T1 Phage-Resistant Cells (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 ml)
Store S.O.C. Medium at 4°C or room temperature.

.
Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid High Copy Plasmid
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL
Show More Show Less
You offer competent cells in Subcloning Efficiency, Library Efficiency and MAX Efficiency. How do these differ?

There are a few exceptions, but in general the difference is in guaranteed transformation efficiency as follows:

Subcloning Efficiency cells are guaranteed to produce at least 1.0 x 10E6 transformants per µg of transformed pUC19 or pUC18 supercoiled plasmid
Library Efficiency cells are guaranteed to produce at least 1.0 x 10E8 transformants per µg pUC19 or pUC18 DNA
MAX Efficiency cells are guaranteed to produce at least 1.0 x 10E9 transformants per µg pUC19 or pUC18 DNA

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.