Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCRX Enhancer System

Catalog No. 11495017
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-495-017 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-495-017 Supplier Invitrogen™ Supplier No. 11495017
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

PCRx Enhancer Solution consists of 10X PCRx Amplification Buffer, 10X PCRx Enhancer Solution, and 50mM magnesium sulfate.

PCRx Enhancer System is used in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates.

PCRX Enhancer System is an optimized buffer and cosolvent system designed for use in conjunction with DNA polymerases to optimize PCR of problematic and/or GC-rich templates. 10X PCRX Enhancer Solution with 10X PCRX Amplification Buffer offers higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostability of DNA polymerases such as Taq DNA polymerase and Platinum Taq DNA Polymerase.

Conventional PCR buffer components can be directly substituted with the 10X PCRX amplification buffer and 50 mM MgSO4 for a more robust amplification and improved product yield in some cases.

Notes

Optimal concentration of 10X PCRX Enhancer Solution will vary depending on GC content, Mg++ concentration, and annealing temperature.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Standard PCR
Quantity 250 Reactions
Includes • 10X PCRx Enhancer Solution (2 x 1 mL)
• 10X PCRx Amplification Buffer (2 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.