Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ III One-Step RT-PCR System with Platinum™ Taq High Fidelity DNA Polymerase

Catalog No. 12574030
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
25 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
12-574-030 25 Reactions
12-574-035 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-574-030 Supplier Invitrogen™ Supplier No. 12574030

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity is designed for convenient end-point detection and analysis of RNA molecules by one-step RT-PCR.

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity is designed for convenient end-point detection and analysis of RNA molecules by one-step RT-PCR. This one-step formulation allows for both cDNA synthesis and PCR amplification to be performed in a single tube using gene-specific primers and target RNAs from either total RNA or mRNA. It also allows for the detection of a wide range of RNA targets (up to 10 kb in length) at variable concentrations (1 pg to 1 μg of total RNA). This system consists of two major components: SuperScript III RT/Platinum Taq High Fidelity enzyme mix and 2X reaction mix.

SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA and can be used to synthesize cDNA from total RNA. Platinum Taq DNA Polymerase High Fidelity is a an enzyme mixture composed of recombinant Taq DNA polymerase, Pyrococcus species GB–D polymerase, and Platinum Taq antibodies, which block polymerase activity at ambient temperatures enabling hot start PCR.

The 2X reaction mix consists of a proprietary buffer system optimized for reverse transcription and PCR amplification, Mg2+ optimized for universal use, deoxyribonucleotide triphosphates, and stabilizers.

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity offers:
• High-fidelity end-point detection and analysis of RNA molecules by RT-PCR
• PCR amplification and cDNA synthesis in a single tube
• Detection of a wide range of RNA targets from 300 bp to 10 kb
• Increased specificity with wide cDNA synthesis temperature range of 45−60°C

Note: For superior one-step RT-PCR performance, we recommend the SuperScript IV One-Step RT-PCR System (Cat. No. 12594025) or SuperScript IV One-Step RT-PCR System with ezDNase (Cat. No. 12595025). The SuperScript IV One-Step RT-PCR System combines the high processivity of SuperScript IV Reverse Transcriptase with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched product yields, specificity, and sensitivity in less time and for a broad target range, even with suboptimally pure RNA samples.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• SuperScript III RT/Platinum Taq High Fidelity Enzyme Mix (25 μL)
• 2X Reaction Mix (1 mL)
• 5 mM Magnesium Sulfate (500 μL)

Store at –10 to –30°C.

Detection Method Gel Electrophoresis
Format One-Step RT-PCR System
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Polymerase Platinum Taq High Fidelity
Reaction Speed 30 min.
Technique 1-Step RT-PCR
Optimal Reaction Temperature 50°C
Quantity 25 rxns
Reverse Transcriptase SuperScript III
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
Final Product Type PCR Amplified cDNA
Hot Start Built-In Hot Start
No. of Reactions 25 Reactions
Reaction Format Premixed Components
Reagent Type Reverse Transcription
Size (Final Product) Up to 10 kb
Starting Material RNA
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.