Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteinase K Solution

Catalog No. 4333793
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-337-93 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-337-93 Supplier Invitrogen™ Supplier No. 4333793
Get 15% Off Sample Preparation Products
Use promo code SAMPREP15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range (optimal activity between 6.5 and 9.5)
  • Under denaturing conditions (e.g., the presence of SDS or urea)
  • In the presence of metal chelating agents (e.g. EDTA)
  • At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and Quality Testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit Definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Purity DNase-Free, RNase-Free
For Use With (Application) Chromatin Biology/RNA Extraction/DNA Extraction
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice
Quantity 5 mL
Product Type Proteinase K
Form Liquid
How do you recommend storing proteinase K?

We recommend storing it at 4°C.

How do I isolate DNA from single bacterial colonies before PCR?

Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.