Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Filter Maxiprep Kit

Catalog No. K210017
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Preps
25 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210017 25 Preps
K210016 10 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K210017 Supplier Invitrogen™ Supplier No. K210017
Add to Cart
Edge
Add to Cart

Includes

The PureLink HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns.

The PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of the highest purity from 25 to 100 mL of overnight bacterial culture.

The PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of the highest purity from 25 to 100 mL of overnight bacterial culture. The PureLink HiPure Plasmid Filter Kits include HiPure Filter Columns, which combine filtration for clarification of bacterial lysate with anion-exchange resin purification of plasmid DNA, in single unit. The resulting plasmid DNA is of quality equivalent to that obtained by purification with two passes through cesium chloride gradients—the most rigorous method for DNA purification.

  • Purify high-quality DNA in Less than two hours for transfections and other applications with no need for additional steps to remove contaminants like RNA, proteins, or endotoxins
  • Additionally, elimination of phenol, chloroform, ethidium bromide, and cesium chloride from protocol minimizes exposure to and disposal of hazardous materials

PureLink HiPure Plasmid Filter Kits provide:

  • Simple protocols—bacterial lysate clarification without centrifugation
  • Fast results save time by purifying plasmid DNA in less than two hours
  • High yields up to 200 μg for midipreps and up to 850 μg for maxipreps
  • Reliable performance, high-quality plasmid DNA purified with these kits is suitable for wide variety of applications

How it works

  • Addition of a HiPure Filter Column to the PureLink HiPure Plasmid Kits provides rapid clearing of the bacterial lysate without the need for centrifugation
  • Lysate filtration cartridge is integrated into DNA-binding column for a combined step of lysate clarification and plasmid DNA binding directly to the anion-exchange resin
  • Single column wash removes impurities such as RNA, proteins, metabolites, and other low molecular weight molecules; the ultrapure plasmid DNA is then eluted with high-salt buffer
  • DNA is then desalted and concentrated with an isopropanol precipitation step, then collected by centrifugation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Column Type Filter Column
Content And Storage The PureLink™ HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns. Store all components at room temperature upon receipt.
Format Column
Isolation Technology Anion Exchange Resin
Scale Maxi
Sample Type Bacterial Culture
Elution Volume 15 mL
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 25 Preps
Plasmid <40kb, Low Copy Plasmid, High Copy Plasmid, BAC
Prep Scale > 200 μg (Large-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid Filter MaxiPrep Kit
Quantity 25 Preps
Purity Transfection Grade
Shipping Condition Room Temperature
Target Plasmid DNA
System Type PureLink™
Test Time 90 min.
Yield 1000 μg
Show More Show Less
I'm getting low/no plasmid DNA after purification using a PureLink HiPure kit, even though there was measurable absorbance. Do you have any suggestions for what I can do?

A common problem encountered with absorbance measurements is turbidity of samples. (This could be caused by residual resin from the column.) If there is insoluble material in the cuvette (not often detected by the naked eye), much of the UV light is not absorbed but scattered, leading to an artificially high UV absorbance reading (at 260 or 280 nm, for example.) If your A260 is high, we recommend that you check the A320 to determine if there is resin in the sample. You can also try to centrifuge or filter (0.2 µm filter) your sample to remove any resin and then recheck the concentration.

I've run out of buffer when using the PureLink HiPure Plasmid Purification Kit (Cat. No. K210018). Can I purchase the buffers separately?

Yes, we would recommend purchasing the PureLink HiPure BAC Buffer Kit (Cat. No. K210018). This kit includes Resuspension Buffer (R3) (250 ml), Lysis Buffer (L7) (250 ml), Precipitation Buffer (N3) (250 ml), and RNase A (20 µg/ml) (5 ml).
You will need to add less RNase A than stated on the bottle label of the R3 buffer in this kit. It says to add 5.6 mL of RNase A. This is the correct amount for the BAC protocol; however, if you are performing standard plasmid isolation, 1.4 mL RNase A should be added.

Plasmid DNA isolated using a PureLink column-based purification kit from an endA+ strain is degraded after a restriction digest. Do you have a suggestion for this?

The HiPure kits should remove all protein from the DNA including endonucleases. For the silica-based PureLink Quick Plasmid Miniprep Kit, we recommend an extra wash with the optional Wash Buffer W10 to remove endonucleases. This solution is not compatible with the HiPure system and should not be used with those kits. Alternatively, heat the eluted DNA in TE for 10 min at 70 degrees C. This should heat-inactivate any contaminating nucleases.

I'm seeing extra bands present after plasmid purification using your PureLink column-based system. What could cause this to happen?

Extra bands can occur when plasmid DNA is nicked and/or permanently denatured. Plasmid DNA that has been nicked (covalently opened) will run slower than supercoiled DNA during electrophoresis. A small amount of this species of DNA is common and is suitable for downstream applications. Permanently denatured DNA will migrate ahead of the supercoiled DNA and may not be suitable for downstream applications. Do not allow the lysis reaction to proceed longer than 5 minutes.

My purified DNA has particles in it after column-based plasmid purification. Any suggestions?

We have seen this on occasion. The particles do not affect quality of the DNA. Remove the particles by performimg a 1 minute centrifugation at 12,000 x g.

My cell lysis solution is cloudy and precipitated when using your column-based plasmid purification kit. How do I redissolve the precipitate?

This is common if the solution gets too cold. If necessary, warm the solution briefly to 37 degrees C until the precipitate is dissolved.

The plasmid I purified using your column-based plasmid purification kit has contaminating genomic DNA present. What should I do?

If contaminating DNA is seen, there may have been problems with the cell lysis and neutralization steps. Perhaps the mixture was not centrifuged appropriately or the sample was mixed too harshly during the lysis step.

The plasmid I purified using your column-based plasmid purification kit has contaminating RNA present. What should I do?

To reduce RNA contamination:

- Increase the volume of Precipitation Buffer (N3) by 10%. (This will reduce the amount of RNA binding to the resin.)
- Increase the salt concentration in the Wash Buffer (W8) from 800 mM NaCl to 860 mM NaCl by adding 0.35 g NaCl per 100 ml Wash Buffer. (This will wash more of the RNA off the resin.)
(NOTE: Each of these changes alone reduces the amount of RNA contamination significantly. Together these changes eliminate virtually all RNA).
- Make sure that the RNase A was added and thoroughly dissolved in the resuspension buffer.
- PureLink HiPure Filter Midi and Maxi Kits may help reduce RNA contamination by clearing bacterial lysate more thoroughly.
- If RNA contamination persists, the RNase A concentration in the resuspension buffer can be increased. It can go as high as 400 µg/mL. This is particularly useful when isolating low-copy plasmids or other single-copy DNA forms such as BACs.
- Contaminating RNA tends to be a little more of a problem when low copy number plasmids are purified due to a large number of cells being processed.

To troubleshoot the bacterial culture:

- The columns may have been overloaded with too many cells. Verify that the culture used was grown to the proper OD value and that the volumes used were within kit specifications. Verify the medium used, as TB or very rich broth could produce a very dense culture, which may cause reduced lysis.
- Verify volumes of cells and buffers added (resuspension with RNAse A, lysis, neutralization steps). Check on amount of RNase added to resuspension buffer. For cultures with high density or high volumes, you could add extra RNase A, to 400 µg/mL as mentioned above.

The plasmid I purified using your column-based plasmid purification kit does not work in my experiment. What can I do?

Please check to see whether the plasmid was ethanol precipitated and washed after elution from the column. Inhibition may occur if there is too much salt and/or if the pH is too high.

I'm getting no or low yields from my column-based plasmid purification experiment. What do you suggest I try?

Here are some suggestions to try:

- Make sure the binding of the plasmid is being done at room temperature (RT). Temperature affects the pH of the binding solution. Make sure all other solutions were also warmed to RT.
- Verify that the centrifugation immediately following the neutralization step was not done at 4 degrees C. If it was, the supernatant MUST be warmed to RT before binding on the column. We find that the DNA binds to the matrix of the columns better if the lysate is at room temperature.
- Low copy number plasmid may have been used. Check plasmid.
- Not all the medium may have been removed at the cell harvesting step, so the pH of the subsequent steps was affected.
- The cell pellet may not have been thoroughly resuspended in the resuspension step.
- Purified DNA may have been overdried after isopropanol precipitation and ethanol wash. Only air-dry the pellet.
- Pellet may have been lost during the isopropanol precipitation and ethanol wash. Be careful at this step, as the pellet tends to be slippery. It is best to pipette off alcohol solutions rather than pour them off.
- When working with BAC DNA (and therefore increasing the NaCl from 0.8 M to 0.9 M in the Wash Buffer W8), an excessive volume of Wash Buffer W8 may have been used. See the BAC prep section in the manual.
- Try elution of DNA with heated elution buffer: For plasmids less than 10 kb, no heating is required. For 10-30 kb, heating (65-70 degrees C) is optional, and may increase elution efficiency by approximately 20%. For plasmids larger than 30 kb, heating is recommended, and may increase elution efficiency by approximately 50%. Perform an additional elution to increase yield by up to 10%.
- If there is some insoluble material in the eluted DNA, it could be resin particles (resin fines). These are inert and can be removed by a centrifugation at 12,000 x g for 1 minute at RT.
- You can also try adding chloramphenicol to the culture at mid-log phase (around OD590 0.8-1.0).

What growth conditions do you recommend for E. coli when isolating plasmid DNA using your PureLink HiPure kits?

We typically recommend growing E. coli up to an optical density of 2.0 at 600 nm or a cell density of approximately 1 x 10^9 cells/mL in LB broth.

Can I isolate plasmid DNA from mammalian cells using your PureLink purification kits?

Yes, please follow our suggested protocol:

1) Lyse mammalian cells with a common mammalian lysis procedure, for instance, alkaline lysis. Please note, be careful to be gentle in this step, as sheared genomic DNA will copurify with plasmid.
2) Precipitate DNA with ethanol or 70-80% isopropanol. This is a crude precipitate.
3) Resuspend in 600 mM NaCl, 100 mM NaOAC, pH adjusted to 5.0.
4) Load onto equilibrated HiPure column.

This should work for plasmids up to 100 kb. Note that mitochondrial DNA will also copurify.

Can I isolate plasmid DNA from gram-positive bacteria using your PureLink purification kits?

1.Lysis conditions vary greatly for gram-positive bacteria, so you would need to start with a specific lysis protocol that is known to work for the particular bacteria you are working with. The lysis materials and conditions in the kit may not work well.

2.After lysis using the specific protocol, precipitate the crude plasmid DNA with either 70% ethanol or isopropanol. Be sure that the genomic DNA is removed (follow precautions in the package insert; do not vortex at any time).

3.Resuspend the resulting nucleic acid pellet in a small volume (1 mL for mini, 10 mL for midi, 24 mL for maxi) of 600 mM NaCl, 100 mM sodium acetate, pH 5.0. Apply this solution to an equilibrated column and continue with the standard protocol.

Can I isolate lambda phage DNA using your PureLink HiPure Mini, Midi, or Maxi Purification Kits?

Yes, please see the detailed protocol below:

1.Equilibrate a column with 2 mL (mini)/10 mL (midi)/30 mL (maxi) of buffer EQ1.
2.Collect the phage lysate (liquid or plate lysis) and determine the exact volume.
3.According to the scale, add 30 µL/100 µL/400 µL of buffer X1 to 10 mL/50 mL/250 mL phage lysate and incubate at 37 degrees C for 30 min.
X1 = 100 mM Tris-HCl (pH 7.5), 300 mM NaCl, 10 mM EDTA, 20 mg/mL RNase A, 6 mg/mL DNase I
4.Mix the nuclease digest from step 3 with 2 mL/ 10 mL/50 mL ice-cold buffer X2 and incubate on ice for 60 min.
X2 = 3 M NaCl, 30% (w/v) polyethyleneglycol (PEG) 6000
5.To collect the phage particles, centrifuge for 10 minutes at greater than 10,000 x g. Discard the supernatant.
6.Resuspend the pelleted phage particles in 1 mL (mini)/3 mL (midi)/9 mL (maxi) buffer X3 with a pipet.
X3 = 100 mM Tris-HCl (pH 8.0), 25 mM EDTA
7.Add 1 mL (mini)/3 mL (midi)/9 mL (maxi) of buffer X4 to the phage suspension. Mix thoroughly by inverting the tube several times and incubate for 10 minutes (mini) or 20 minutes (midi and maxi) at 70 degrees C to lyse the phage particles.
X4 = 4% (w/v) SDS
8.Add 1 mL (mini)/3 mL (midi)/9 mL (maxi) of buffer X5 to the lysate, mix thoroughly by inverting, and centrifuge for 10 minutes at room temperature and ?13,000 x g. Collect the supernatant without taking too many particles and apply it directly onto the equilibrated column (see step 1). Allow the lysate to enter the resin by gravity flow.
X5 = 3.0 M potassium acetate (pH 5.5 with acetic acid)
9.Wash the column with 2 x 2.5 mL (mini), 2 x 10 mL (midi), or 1 x 60 mL (maxi) of buffer W8.
10.Elute the lambda DNA from the column with 0.9 mL (mini)/5 mL (midi)/15 mL (maxi) of buffer X6 and precipitate the DNA by adding 0.7 volumes of isopropanol, previously equilibrated to room temperature.
X6 = 100 mM sodium acetate (pH 5.0 with acetic acid), 1,500 mM NaCl
11.Centrifuge the DNA for 30 minutes at ?13,000 x g at 4 degrees C. Because lambda DNA is very sticky, it will spread over the whole wall of the centrifuge tube if a fixed angle rotor is used. Therefore, we recommending the use of a swinging bucket rotor (i.e. HB-4 or HB-6 for Sorvall centrifuges), or, if such a rotor is not available, using centrifuge tubes (i.e., Corex) siliconized with dimethyldichlorosilane.

After centrifugation, was the lambda DNA with 80% ethanol and dry it briefly. Dissolve the lambda DNA in a suitable amount of TE or 10 mM Tris buffer (pH 8.0).

What is the maximum plasmid size for plasmid purification using the PureLink HiPure kits?

The PureLink HiPure Mini, Midi, and Maxi Kits can purify plasmids up to 200 kb, while the PureLink HiPure Mega and Giga Kits can purify plasmids up to 150 kB.

What are typical yields for your PureLink HiPure Plasmid Kits?

Miniprep
Overnight bacterial culture volume: 1-3 mL
Approximate yield for high copy plasmids: ≤30 µg

Midiprep
Overnight bacterial culture volume: 15-25 mL
Approximate yield for high copy plasmids: 100-350 µg

Maxiprep
Overnight bacterial culture volume: 100-200 mL
Approximate yield for high copy plasmids: 500-850 µg

Can I decrease the volume of elution buffer I'm using with the PureLink Plasmid Purification Kits in order to increase yield?

We do not recommend decreasing the volume of elution buffer, as this will cause yield to drop. You can try to perform an additional elution to increase yield.

Can I elute my plasmid from PureLink columns with water instead of TE?

For any silica columns, elution with water is generally possible. However, a buffer is preferred for stability and accuracy of absorbance readings, as pure water can have a very low pH (4 - 5).

Can the PureLink HiPure plasmid kits isolate bacterial artificial chromosome (BAC) DNA, bacmid DNA, cosmid DNA, or M13 ssDNA?

Yes, the PureLink HiPure Plasmid kits can isolate BAC DNA, bacmid DNA, cosmid DNA, or M13 ssDNA. See the manual for a detailed protocol.

What's the difference between PureLink HiPure Plasmid Kits, PureLink HiPure Plasmid Filter Kits, and PureLink HiPure Plasmid FP kits?

All kits termed ‘HiPure' use anion exchange resin columns to isolate the highest quality plasmid DNA, suitable for transfection. Filter kits include a filter to clear bacterial lysate without centrifugation, while the HiPure FP kits include a filter and precipitator to eliminate the need for centrifugation at either the bacterial lysate clearing or DNA precipitation step.

Do you offer a vacuum manifold for purification of nucleic acids?

Yes, we offer our EveryPrep Universal Vacuum Manifold (Cat. No. K2111-01), which allows for direct elution from the manifold using our ChargeSwitch Pro Filter Plasmid Mini, Midi, and Maxi Kits.

What are endotoxins and why are their levels important for transfection?

Endotoxins are typically any cell-associated bacterial toxins that are part of the outer surface of the cell wall of gram-negative bacteria. Endotoxins can influence cell growth, cell differentiation, contractility, and protein expression in mammalian cells. Endotoxins are released during bacterial lysis, and they can subsequently reduce transfection efficiency and protein expression levels. Please review the following article for more information about endotoxins: Butash KA et al. (2000) Reexamination of the effect of endotoxin on cell proliferation and transfection efficiency. Biotechniques 29(3): 610-614, 616, 618-619.

I need to purify a cloned plasmid, which I will be transfecting into a cell line for protein expression. I have both silica and anion exchange columns available. Which would you recommend as the better option for my application?

Anion exchange purification is recommended for higher purity and lower endotoxin levels. Silica-based purification is not optimal for transfection, as there is a higher level of endotoxins and impurities. Larger plasmids also work better with anion exchange columns.

What is the best way to determine the quality/purity of my plasmid DNA?

The ratio of absorbance at 260 nm to the absorbance at 280 nm (A260/A280) is typically used to measure purity of the sample. For DNA, the ideal A260/A280 ratio is 1.8, but it can be in the range of 1.7 - 1.9. The A260/A230 ratio is also used to determine if contamination is present. For DNA, the ideal A260/A230 ratio is between 1.8 and 2.0. DNA purity can also be examined by gel analysis. For plasmid DNA, look for a strong, single band (perhaps with a few extra bands representing multimers of the desired molecule). For genomic DNA, look for high average fragment sizes.

Can I use water instead of TE buffer to elute plasmid DNA from the PureLink HiPure Plasmid kits or Precipitators?

Yes, but water of low pH or poor quality may cause a decrease in yield and DNA quality. The TE buffer provided with the PureLink HiPure Plasmid Purification Kits contains 0.1 mM EDTA, which is compatible with most subsequent reactions.

Can the PureLink HiPure Filter and Precipitator kits be used for large plasmids?

At this time, for purification of large plasmids (>50 kb), we recommend using the standard PureLink HiPure Plasmid Kits (without the HiPure Filter or Precipitator) for purification of large plasmids. There are protocols in PureLink HiPure Plasmid DNA Purification manual for Bacmid, BAC and cosmid.

What are the compositions of the buffers included in the PureLink HiPure mini, midi, and maxi prep kits?

The buffer compositions are as follows:
Resuspension Buffer (R3): 50 mM Tris-HCl (pH 8.0), 10 mM EDTA
RNase A: (20 mg/mL in R3)
Lysis Buffer (L7): 200 mM NaOH, 1% SDS (w/v)
Precipitation Buffer (N3): 3.1 M potassium acetate (pH 5.5)
Equilibration Buffer (EQ1): 600 mM NaCl, 100 mM sodium acetate(pH 5.0), 0.15% Triton X-100 (v/v)
Wash Buffer (W8): 800 mM NaCl, 100 mM sodium acetate (pH 5.0)
Elution Buffer (E4): 1.25 M NaCl, 100 mM Tris-HCl (pH 8.5)

Please note that these buffer composition data apply to the PureLink HiPure Plasmid Mini, Midi, Maxi, Mega, and Giga kits.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.